Skip to Content
Merck
All Photos(1)

Key Documents

C6554

Sigma-Aldrich

Casein from bovine milk

BioReagent, suitable for insect cell culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

bovine milk

product line

BioReagent

form

powder

technique(s)

cell culture | insect: suitable

mp

280 °C (dec.) (lit.)

solubility

1 M NaOH: 50 mg/mL, turbid, faintly yellow

UniProt accession no.

storage temp.

room temp

SMILES string

[P](=O)(OCC(NC(=O)C(NC(=O)C(N)Cc1ccccc1)CCC(=O)N)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(C(O)C)C(=O)NC(CCC(=O)O)C(=O)NC(CC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CC(C)C)C(=O)NC(CCC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC(C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Numerous experimental applications including use as a blocking agent in immunochemistry, recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.
Partial gastrointestinal digestion of casein is a rich source of bioactive peptides, such as β-casomorphin. However, bovine casein is not homogeneous; variants A1 and B do lead to production of β-casomorphin 7 production, while A2 does not.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cynthia Martinez-Liu et al.
Biosensors, 12(7) (2022-07-28)
Flavivirus detection in humans and mosquito reservoirs has been an important issue since it can cause a variety of illnesses and could represent a health problem in geographical zones where the vector is endemic. In this work, we designed and
Krisztina Manzano-Szalai et al.
PloS one, 11(3), e0150819-e0150819 (2016-03-11)
In highly sensitized patients, the encounter with a specific allergen from food, insect stings or medications may rapidly induce systemic anaphylaxis with potentially lethal symptoms. Countless animal models of anaphylaxis, most often in BALB/c mice, were established to understand the
Yeonho Chang et al.
Experimental & molecular medicine, 53(3), 384-392 (2021-03-04)
Single-molecule localization microscopy (SMLM) has allowed the observation of various molecular structures in cells beyond the diffraction limit using organic dyes. In principle, the SMLM resolution depends on the precision of photoswitching fluorophore localization, which is inversely correlated with the
K Benschop et al.
Journal of clinical microbiology, 46(12), 3965-3970 (2008-10-24)
Human parechoviruses (HPeV) are widespread pathogens belonging to the Picornavirus family. Six genotypes, which have predominantly been isolated from children, are known. Data on prevalence of HPeV genotypes are generally based on cell culture, which may underestimate the prevalence of
Losee L Ling et al.
Nature, 517(7535), 455-459 (2015-01-07)
Antibiotic resistance is spreading faster than the introduction of new compounds into clinical practice, causing a public health crisis. Most antibiotics were produced by screening soil microorganisms, but this limited resource of cultivable bacteria was overmined by the 1960s. Synthetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service