Skip to Content
Merck

Skip To

L6010

α-Lactalbumin from bovine milk

Type III, calcium depleted, ≥85% (PAGE), lyophilized powder

Synonym(s):

Bos d 4, Lactose synthase B protein, alpha-lactalbumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.61
MDL number:
Biological source:
bovine milk
Assay:
≥85% (PAGE)
Form:
lyophilized powder
Technique(s):
indirect ELISA: suitable
Impurities:
cation traces, tested
Concentration:
≥85 % protein
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine milk

Quality Segment

type

Type III

assay

≥85% (PAGE)

form

lyophilized powder

quality

calcium depleted

mol wt

14,178 Da by calculation

concentration

≥85 % protein

technique(s)

indirect ELISA: suitable

impurities

cation traces, tested

solubility

H2O: soluble 10 mg/mL, clear to slightly hazy, colorless to faintly yellow

cation traces

Ca: ≤0.3 mol/mol

UniProt accession no.

storage temp.

−20°C

Gene Information

cow ... LALBA(281894)

General description

may contain traces of (NH4)2SO4 and sodium phosphateL6010

Application

α-Lactalbumin from bovine milk has been used:
  • in indirect enzyme-linked immunosorbent assay (ELISA) and competitive ELISA methods
  • in binding of Hsp90 and HSJ1b analysis[1]
  • to inhibit the lysozyme CAP-RAST assay

Biochem/physiol Actions

α-Lactalbumin is the cheif protein in human milk. It consists of a single polypeptide chain with 8 cysteines which form disulfide bridges. α-Lactalbumin binds several metal ions, including calcium, which is thought to play a role in the regeneration of native α-lactalbumin from the reduced, denatured form.[2] α-Lactalbumin also has a distinct zinc binding site that is thought to play a role in the binding of the lactose synthase complex.[3][4]The mature protein consists of 123 amino acid residues (14 kD), and it has a three-dimensional structure with 1.7 Angstrom resolution, demonstrating four α-helices and a triple stranded antiparallel β-sheet.[5]
Alters the substrate specificity of galactosyltransferase to increase the rate of lactose formation; the complex of galactosyltransferase and α-lactalbumin is called lactose synthase.
Alters the substrate specificity of galactosyltransferase to increase the rate of lactose formation; the complex of galactosyltransferase and α-lactalbumin is called lactose synthase. Site-directed mutagenesis of Asp87 or Asp88 to Ala completely abolishes the strong calcium binding affinity and reduces the stimulation of lactose synthase to <3.5% of the maximal rate.

Analysis Note

May contain traces of (NH4)2SO4 and sodium phosphate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L5385L7269L6385
technique(s)

indirect ELISA: suitable

technique(s)

cell culture | mammalian: suitable, electrophoresis: suitable

technique(s)

cell migration: suitable

technique(s)

electrophoresis: suitable

biological source

bovine milk

biological source

bovine milk

biological source

human milk

biological source

bovine milk

assay

≥85% (PAGE)

assay

≥85% (PAGE)

assay

≥95% (SDS-PAGE)

assay

-

Quality Level

300

Quality Level

300

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

powder

impurities

cation traces, tested

impurities

calcium, tested

impurities

-

impurities

-


Still not finding the right product?

Explore all of our products under α-Lactalbumin from bovine milk


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters