Skip to Content
Merck
All Photos(2)

Key Documents

A8292

Sigma-Aldrich

Acetobromo-α-D-glucuronic acid methyl ester

≥93% (GC)

Synonym(s):

(2,3,4-Tri-O-acetyl-α-D-glucopyranosyl bromide)uronic acid methyl ester, Bromo-2,3,4-tri-O-acetyl-α-D-glucopyranuronic acid methyl ester, Methyl acetobromo-α-D-glucuronate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17BrO9
CAS Number:
Molecular Weight:
397.17
Beilstein:
96281
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥93% (GC)

form

powder

optical activity

[α]/D 188 to 198 °, c = 1% (w/v) in ethyl acetate

color

white

mp

80-110 °C (lit.)

solubility

ethyl acetate: 50 mg/mL, clear, colorless to faintly yellow

shipped in

dry ice

storage temp.

−20°C

SMILES string

COC(=O)[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O

InChI

1S/C13H17BrO9/c1-5(15)20-8-9(21-6(2)16)11(13(18)19-4)23-12(14)10(8)22-7(3)17/h8-12H,1-4H3/t8-,9-,10+,11-,12-/m0/s1

InChI key

GWTNLHGTLIBHHZ-SVNGYHJRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetobromo-α-D-glucuronic acid methyl ester is used for the synthesis of HMR1098-S-Glucuronide Methyl Ester, a new K-ATP-blocking agent being developed as a drug for prevention of sudden cardiac death.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian T Ethell et al.
Drug metabolism and disposition: the biological fate of chemicals, 31(8), 1027-1034 (2003-07-18)
HMR1098, a novel KATP-blocking agent, is metabolized to form an S-glucuronide in rat and dog bile. Synthesis of the S-glucuronide metabolite was studied in human liver and kidney microsomes. Recombinant UPD-glucuronosyltransferases (UGTs) were screened for activity, and kinetic analysis was
John C Quindry et al.
American journal of physiology. Heart and circulatory physiology, 299(1), H175-H183 (2010-05-04)
The mechanisms responsible for anti-arrhythmic protection during ischemia-reperfusion (IR) in exercised hearts are not fully understood. The purpose of this investigation was to examine whether the ATP-sensitive potassium channels in the mitochondria (mito K(ATP)) and sarcolemma (sarc K(ATP)) provide anti-arrhythmic
Katarzyna Choma et al.
Journal of bioenergetics and biomembranes, 41(4), 323-334 (2009-10-13)
Mitochondrial potassium channels in the brain have been suggested to have an important role in neuroprotection. The single channel activity of mitochondrial potassium channels was measured after reconstitution of the purified inner membrane from rat brain mitochondria into a planar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service