Skip to Content
Merck
All Photos(1)

Key Documents

P50404

Sigma-Aldrich

1,3-Propanediol

98%

Synonym(s):

1,3-Dihydroxypropane, Trimethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)3OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
969155
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.8 mmHg ( 20 °C)
9.8 mmHg ( 100 °C)

Quality Level

Assay

98%

refractive index

n20/D 1.440 (lit.)

bp

214 °C/760 mmHg (lit.)

mp

−27 °C (lit.)

density

1.053 g/mL at 25 °C (lit.)

SMILES string

OCCCO[H]

InChI

1S/C3H8O2/c4-2-1-3-5/h4-5H,1-3H2

InChI key

YPFDHNVEDLHUCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Propanediol can be used as:
  • A solvent as well as stabilizer in the preparation of thin films.
  • A monomer to synthesize various polyesters such as poly(propylene terephthalate) (PPT), poly(propylene isophthalate) (PPI), and poly(propylene naphthalate) by polycondensation reaction with terephthalic, isophthalic, and 2,6-naphthalenedicarboxylic acid.
  • A reactant to prepare 7,8-benzoquinoline by IrCl3 catalyzed cyclization reaction with 1-naphthylamine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>210.2 °F - closed cup

Flash Point(C)

> 99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Materials Science Letters, 12, 1221-1221 (1993)
Effect of different calcination temperatures and post annealing on the properties of 1, 3 propanediol based Sol-Gel (Na0. 5K0. 5) NbO3 (NKN) thin films
Wiegand S, et al.
Journal of alloys and compounds, 548, 38-45 (2013)
N-heterocyclization of naphthylamines with 1,2- and 1,3-diols catalyzed by an iridium chloride/BINAP system
Hiroomi Aramoto, et al.
The Journal of Organic Chemistry, 74(2), 628-623 (2009)
Stud. Nat. Prod. Chem., 13, 53-53 (1993)
Synthesis, thermal characterization, and tensile properties of alipharomatic polyesters derived from 1, 3-propanediol and terephthalic, isophthalic, and 2, 6-naphthalenedicarboxylic acid
Roupakias CP, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 43(17), 3998-4011 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service