
Acetobromo-α-D-glucuronic acid methyl ester

Synonym(s):
(2,3,4-Tri-O-acetyl-α-D-glucopyranosyl bromide)uronic acid methyl ester, Bromo-2,3,4-tri-O-acetyl-α-D-glucopyranuronic acid methyl ester, Methyl acetobromo-α-D-glucuronate
Empirical Formula (Hill Notation):
C13H17BrO9
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing