Skip to Content
Merck
All Photos(4)

Key Documents

A6106

Sigma-Aldrich

Aprotinin

BioUltra, 3-8 TIU/mg solid, powder, ≥98% (SDS-PAGE), suitable for inhibition assay

Synonym(s):

BPTI, Bovine pancreatic trypsin inhibitor, Trasylol, Trypsin inhibitor (basic)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C284H432N84O79S7
CAS Number:
Molecular Weight:
6511.44
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.77

Product Name

Aprotinin from bovine lung, BioUltra, 3-8 TIU/mg solid, ≥98% (SDS-PAGE)

biological source

bovine lung

product line

BioUltra

Assay

≥98% (SDS-PAGE)

form

powder

specific activity

3-8 TIU/mg solid

mol wt

~6,500

technique(s)

inhibition assay: suitable

solubility

H2O: 5 mg/mL, clear to very slightly hazy

UniProt accession no.

storage temp.

2-8°C

SMILES string

S1SCC2NC(=O)CNC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C5NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C9N(CCC9)C(=O)CNC(=O)C(NC(=O)C(NC(=O)C%11N(CCC%11)C(=O

InChI

1S/C284H432N84O79S7/c1-21-144(9)222-271(439)337-174(68-46-105-309-282(300)301)239(407)340-187(120-160-77-85-164(374)86-78-160)251(419)341-185(116-156-55-29-24-30-56-156)250(418)342-188(121-161-79-87-165(375)88-80-161)252(420)346-191(123-208(291)378)246(41

InChI key

ZPNFWUPYTFPOJU-UHFFFAOYSA-N

Gene Information

cow ... PTI(404172)

Looking for similar products? Visit Product Comparison Guide

Application

Aprotinin is largely used as an inhibitor of trypsin.

Biochem/physiol Actions

Aprotinin is a competitive serine protease inhibitor that forms stable complexes with and blocks the active sites of enzyme. This binding is reversible, and most aprotinin-protease complexes will dissociate at extreme pH levels >10 or <3. Structurally, Aprotinin is a monomeric globular protein derived from bovine lung that consists of 58 amino acids, arranged in a single polypeptide chain with three crosslinking disulfide bridges.

Unit Definition

One Trypsin Inhibitor Unit (TIU) will decrease the activity of two trypsin units by 50%, where one trypsin unit will hydrolyze 1.0 μmole of N-alpha-benzoyl-DL-arginine p-nitroanilide per minute at pH 7.8 and 25°C. Another commonly used unit is the KIU, with 1 TIU = 1,300 KIU.
One trypsin inhibitor unit (TIU) will decrease the activity of 2 trypsin units by 50% where one trypsin unit will hydrolyze 1.0 μmole of N-α-benzoyl-DL-arginine p-nitroanilide (BAPNA) per min at pH 7.8 at 25 °C.

Preparation Note

A further purification of A1153 to yield a product with slightly higher activity and purity

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luciana Mateus Gonçalves et al.
Frontiers in endocrinology, 11, 602519-602519 (2021-02-02)
Pancreatic islets are clusters of endocrine cells that secrete different hormones to regulate blood glucose levels. Efficient hormone secretion requires a close interaction of endocrine cells with their vascular system. Islets receive blood through feeding arteriole(s) that branch into capillaries
Cecilia B Levandowski et al.
PLoS biology, 19(8), e3001364-e3001364 (2021-08-06)
The naturally occurring Δ40p53 isoform heterotetramerizes with wild-type p53 (WTp53) to regulate development, aging, and stress responses. How Δ40p53 alters WTp53 function remains enigmatic because their co-expression causes tetramer heterogeneity. We circumvented this issue with a well-tested strategy that expressed
Renate Gehwolf et al.
Methods in molecular biology (Clifton, N.J.), 2045, 155-165 (2019-02-09)
Tendons harbor various cell populations, including cells displaying classical adult mesenchymal stromal cell criteria. Previous studies have shown that a tenogenic phenotype is more effectively maintained in a 3D cell culture model under mechanical load. This chapter describes a method
Luming Wan et al.
Nature metabolism, 4(1), 29-43 (2022-01-08)
Severe cases of infection with severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) are associated with elevated blood glucose levels and metabolic complications. However, the molecular mechanisms for how SARS-CoV-2 infection alters glycometabolic control are incompletely understood. Here, we connect the
Christian M Cohrs et al.
Cell reports, 31(1), 107469-107469 (2020-04-09)
Type 2 diabetes is characterized by peripheral insulin resistance and insufficient insulin release from pancreatic islet β cells. However, the role and sequence of β cell dysfunction and mass loss for reduced insulin levels in type 2 diabetes pathogenesis are

Articles

While aprotinin and bovine pancreatic trypsin inhibitor (BPTI) are the same protein sequence, the term aprotinin is typically used when describing the protein derived from bovine lung.

Protocols

Objective: To standardize a procedure for the enzymatic assay of Aprotinin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service