Skip to Content

A3428

Aprotinin

3-8 TIU/mg solid, lyophilized powder, suitable for cell culture

Synonym(s):

BPTI, Bovine pancreatic trypsin inhibitor, Trasylol, Trypsin inhibitor (basic)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C284H432N84O79S7
CAS Number:
Molecular Weight:
6511.44
UNSPSC Code:
12352200
eCl@ss:
32190102
NACRES:
NA.32
EC Number:
232-994-9
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid, BioReagent, suitable for cell culture

biological source

bovine lung

Quality Segment

product line

BioReagent

form

lyophilized powder

specific activity

3-8 TIU/mg solid

mol wt

~6,500

technique(s)

cell culture | mammalian: suitable

input

mammalian sample(s)

solubility

H2O: 5 mg/mL

UniProt accession no.

storage temp.

2-8°C

SMILES string

S1SCC2NC(=O)CNC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C5NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C9N(CCC9)C(=O)CNC(=O)C(NC(=O)C(NC(=O)C%11N(CCC%11)C(=O

InChI key

ZPNFWUPYTFPOJU-UHFFFAOYSA-N

Gene Information

cow ... PTI(404172)

Application

Aprotinin from bovine lung has been used in insulin and glucagon-like peptide-1 assay. It has also been used a in western blotting.
Aprotinin is largely used as an inhibitor of trypsin.

Biochem/physiol Actions

Aprotinin is a competitive serine protease inhibitor that forms stable complexes with and blocks the active sites of enzyme. This binding is reversible, and most aprotinin-protease complexes will dissociate at extreme pH levels >10 or <3. Structurally, Aprotinin is a monomeric globular protein derived from bovine lung that consists of 58 amino acids, arranged in a single polypeptide chain with three crosslinking disulfide bridges.
Aprotinin is a natural polypeptide of 58 amino acids. It possesses hemostatic/antiplasmin action and helps to reduce bleeding during surgeries. This prevents the necessity of blood transfusion.

Preparation Note

This product is a lyophilized powder that has been cell culture tested. It has an activity of 3-7 TIU/mg.

Other Notes

One Trypsin Inhibitor Unit (TIU) will decrease the activity of two trypsin units by 50%, where one trypsin unit will hydrolyze 1.0 μmole of N-alpha-benzoyl-DL-arginine p-nitroanilide per minute at pH 7.8 and 25°C. Another commonly used unit is the KIU, with 1 TIU = 1,300 KIU.
When benzoyl-L-arginine ethyl ester (BAEE) is used as substrate one BAPNA unit is approx. 45BAEE μmolar units at pH8.0 at 25°C or approx. 9,000 BAEE A253 units at pH7.6 at 25°C.


Still not finding the right product?

Explore all of our products under Aprotinin from bovine lung


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library