Skip to Content
Merck
All Photos(5)

Documents

A5503

Sigma-Aldrich

Albumin from chicken egg white

lyophilized powder, ≥98% (agarose gel electrophoresis)

Synonym(s):

Albumin from hen egg white, Ovalbumin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201
NACRES:
NA.25

biological source

chicken egg white

type

Grade V

Assay

≥98% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

44287 Da by calculation

purified by

crystallization

technique(s)

bioconjugation: suitable

solubility

H2O: soluble 40 mg/mL

UniProt accession no.

storage temp.

2-8°C

Gene Information

chicken ... OVAL(396058)

Looking for similar products? Visit Product Comparison Guide

General description

Chicken egg albumin, or ovalbumin, is the major protein constituent of egg whites. Chicken egg albumin is a phosphorylated glycoprotein. From the amino acid sequence, the peptide portion of ovalbumin consists of 385 residues and has a molecular weight of 42.7 kDa.

Application

Ovalbumin is useful as a marker (∼45 kDa) for protein electrophoresis. Ovalbumin has been used to produce an animal model of asthma. Ovalbumin was used as a medium for cell incubation in EliCell assay for the detection of released cytokines from eosinophils.
Studies to test the effects of localization on DNA immunizations use ovalbumin in antibody assays.
Ovalbumin has been used as an induction agent in model studies of atopic dermatitis.
A phosphorylated glycoprotein which is useful as a marker (~45 kDa) for protein electrophoresis. Has been used to produce an animal model of asthma.

Biochem/physiol Actions

Differential scanning calorimetry indicates that chicken egg albumin denatures at 84 °C. Ovalbumin can be used as a carrier protein to conjugate to synthetic peptides for use as an immunogen.

Physical form

Essentially salt free

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Boyle et al.
International immunology, 9(12), 1897-1906 (1998-02-18)
We examined the role of the cellular localization of antigen on the immune response after DNA immunization of mice with three forms of ovalbumin (OVA). DNA encoding OVA which was secreted (sOVA) generated 10- to 100-fold higher IgG responses with
A D Nisbet et al.
European journal of biochemistry, 115(2), 335-345 (1981-04-01)
The complete amino acid sequence of hen ovalbumin, comprising 385 residues, has been determined. The sequence was deduced from the 17 cyanogen bromide fragments and from peptides derived by digestion with a number of proteolytic enzymes. The molecular weight of
A differential scanning calorimetric study of conversion of ovalbumin to S-ovalbumin in eggs.
J W Donovan et al.
Journal of the science of food and agriculture, 27(2), 197-204 (1976-02-01)
Joseph Khoory et al.
PloS one, 11(1), e0141206-e0141206 (2016-01-20)
Acute, inflammatory conditions associated with dysregulated complement activation are characterized by significant increases in blood concentration of reactive oxygen species (ROS) and ATP. The mechanisms by which these molecules arise are not fully understood. In this study, using luminometric- and
Ana Raquel Maceiras et al.
Nature communications, 8, 15067-15067 (2017-04-22)
Immunization leads to the formation of germinal centres (GCs) that contain both T follicular helper (Tfh) and T follicular regulatory (Tfr) cells. Whether T-cell receptor (TCR) specificity defines the differential functions of Tfh and Tfr cells is unclear. Here we

Articles

For use as a marker in SDS-PAGE; Albumin from chicken egg white, For use as a marker in SDS-PAGE; L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service