Skip to Content

A3886

Aprotinin

powder

Synonym(s):

BPTI, Bovine pancreatic trypsin inhibitor, Trasylol, Trypsin inhibitor (basic)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C284H432N84O79S7
CAS Number:
Molecular Weight:
6511.44
UNSPSC Code:
12352200
NACRES:
NA.77
EC Number:
232-994-9
MDL number:
eCl@ss:
32190102
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Aprotinin from bovine lung,

biological source

bovine lung

Quality Segment

form

powder

mol wt

~6,500

packaging

vial of 15 mg

input

mammalian sample(s)

solubility

glycerol: 3 mg/mL, clear, colorless (equilibration buffer containing 5% glycerol)

UniProt accession no.

storage temp.

−20°C

SMILES string

S1SCC2NC(=O)CNC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C5NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C9N(CCC9)C(=O)CNC(=O)C(NC(=O)C(NC(=O)C%11N(CCC%11)C(=O

InChI key

ZPNFWUPYTFPOJU-UHFFFAOYSA-N

Gene Information

cow ... PTI(404172)

Application

Aprotinin is largely used as an inhibitor of trypsin.
Gel filtration molecular weight marker

Biochem/physiol Actions

Aprotinin is a competitive serine protease inhibitor that forms stable complexes with and blocks the active sites of enzyme. This binding is reversible, and most aprotinin-protease complexes will dissociate at extreme pH levels >10 or <3. Structurally, Aprotinin is a monomeric globular protein derived from bovine lung that consists of 58 amino acids, arranged in a single polypeptide chain with three crosslinking disulfide bridges.

Other Notes

One Trypsin Inhibitor Unit (TIU) will decrease the activity of two trypsin units by 50%, where one trypsin unit will hydrolyze 1.0 μmole of N-alpha-benzoyl-DL-arginine p-nitroanilide per minute at pH 7.8 and 25°C. Another commonly used unit is the KIU, with 1 TIU = 1,300 KIU.


Still not finding the right product?

Explore all of our products under Aprotinin from bovine lung


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library