Skip to Content
Merck
All Photos(3)

Key Documents

A2737

Sigma-Aldrich

Atractylenolide I

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18O2
CAS Number:
Molecular Weight:
230.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder or crystals

solubility

methanol: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

C=C1CCC[C@@](C=C2O3)(C)[C@@]1([H])CC2=C(C)C3=O

InChI

1S/C15H18O2/c1-9-5-4-6-15(3)8-13-11(7-12(9)15)10(2)14(16)17-13/h8,12H,1,4-7H2,2-3H3/t12-,15+/m0/s1

InChI key

ZTVSGQPHMUYCRS-SWLSCSKDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atractylenolide I is one of the active ingredients of Rhizoma Atractylodes macrocephala which is obtained from the dried root and stem of Atractylodes Macrocephala Koidz. It is a sesquiterpene compound of hydrophobic nature.

Application

Atractylenolide I is a phytochemical that may be used to study its anti-inflammatory and anti-angiogenesis activities. Atractylenolide I may be used and studied as a postitive modulator of GABA-induced chloride currents I(GABA) and as an inhibitor of aromatases. Atractylenolide I may be used as a reference material in assays to detect its presence in plant root extracts and biological milieu such as plasma.

Biochem/physiol Actions

Atractylenolide I has been reported to have an anti-inflammatory activity due to its inhibitory effect on tumor necrosis factor-α (TNF-α) and the production of nitric oxide (NO). It also shows anti-cancer nature, thereby being used in the management of gastric cancer cachexia symptoms. Additionally, it has anti-angiogenic, pro-oxidative and cytotoxic characteristics.
Phytochemical from Traditional Chinese Medicine herbal preparations. Atractylenolide I is an anti-inflammatory that is reported to inhibit angiogenesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

339.6 °F

Flash Point(C)

170.87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Ming Huang et al.
Scientific reports, 4, 3840-3840 (2014-01-24)
Paclitaxel, a known TLR4 ligand, leads to activation of TLR4/MyD88-dependent pathway that mediates chemoresistance and tumor progression in epithelial ovarian carcinoma (EOC). Atractylenolide-I (AO-I), a novel TLR4-antagonizing agent, inhibits TLR4 signaling by interfering with the binding of LPS or paclitaxel
Yujuan Li et al.
Journal of pharmaceutical and biomedical analysis, 58, 172-176 (2011-10-22)
A new high-performance liquid chromatography/tandem mass spectrometry (LC-MS/MS) was developed for quantitative analysis of atractylenolide I in rat plasma using buspirone as internal standard (I.S.). Rat plasma samples were deproteined with methanol and acetonitrile (1:1, v/v). Atractylenolide I and I.S.
Hai Jiang et al.
Molecules (Basel, Switzerland), 16(4), 3146-3151 (2011-04-16)
Ten compounds were isolated from the dichloromethane extract of Atractylodes macrocephala and their aromatase inhibiting activities were tested using an in vitro fluorescent-based aromatase assay. The results indicated that atractylenolide I, atractylenolide II and atractylenolide III had inhibition ratios of
Yi Liu et al.
Evidence-based complementary and alternative medicine : eCAM, 5(3), 337-344 (2008-10-03)
We determined the therapeutic efficacy of atractylenolide I (ATR), extracted from largehead atractylodes rhizome, in managing gastric cancer cachexia (GCC), and interpreted its probable pharmacological mechanism via investigating tumor necrosis factor alpha (TNF-alpha), interleukin-1 (IL-1), interleukin-6 (IL-6) and proteolysis-inducing factor
Hyun Young Park et al.
Planta medica, 77(13), 1528-1530 (2011-02-25)
The roots of Cyathula officinalis Kuan are widely used in Chinese medicine for the treatment of inflammatory disorders. Here, the ability of C. officinalis Kuan to downregulate matrix metalloproteinase (MMP)-13 was examined since MMP-13 is an important enzyme for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service