Skip to Content
Merck
All Photos(1)

Key Documents

SML0415

Sigma-Aldrich

Alantolactone

≥98% (HPLC)

Synonym(s):

(+)-Alantolactone., 3aR,5S,8aR,9aR)-3a,5,6,7,8,8a,9,9a-Octahydro-5,8a-dimethyl-3-methylene-naphtho[2,3-b]furan-2(3H)-one, Alant camphor, Elecampane camphor, Eupatal, Inula camphor, NSC 333843, NSC 93131

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O2
CAS Number:
Molecular Weight:
232.32
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

−20°C

SMILES string

C[C@H]1CCC[C@]2(C)C[C@H]3OC(=O)C(=C)[C@H]3C=C12

InChI

1S/C15H20O2/c1-9-5-4-6-15(3)8-13-11(7-12(9)15)10(2)14(16)17-13/h7,9,11,13H,2,4-6,8H2,1,3H3/t9-,11+,13+,15+/m0/s1

InChI key

PXOYOCNNSUAQNS-AGNJHWRGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Alantolactone is a compound produced by Inula helenium.[1] Alantolactone has antibacterial, antifungal, anti-inflammatory and anticancer properties. It suppresses signal transducer and activator of transcription 3 (STAT3) activation in liver and breast cancer cells. Alantolactone induces apoptosis in A549 lung cancer cells.[2]

Application

Alantolactone has been used:
  • in cell viability assay[1]
  • to examine the percentage of multicentrosomal mitosis in low and high passage human embryonic stem cells (hESC)[3]
  • in assessment of degranulation[4]

Biochem/physiol Actions

Alantolactone is a sesquiterpene lactone disrupts the Cripto-1/ActRII complexes, resulting in an induction of activin/SMAD3 signaling. Alantolactone inhibits proliferation in cancer cells with no affect on normal cells.
Alantolactone is an inducer of activin/SMAD3 signaling.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alantolactone induces apoptosis, promotes STAT3 glutathionylation and enhances chemosensitivity of A549 lung adenocarcinoma cells to doxorubicin via oxidative stress
Maryam A, et al.
Scientific Reports, 7(1), 6242-6242 (2017)
Jaemoo Chun et al.
International immunopharmacology, 14(4), 375-383 (2012-09-04)
Several sesquiterpene lactones are the active components of several medicinal plants and have been demonstrated to perform various pharmacological functions. In this study, we investigated the anti-inflammatory effects of alantolactone, a sesquiterpene lactone isolated from the root of Aucklandia lappa
D E Chapman et al.
Biochemical pharmacology, 38(22), 3913-3923 (1989-11-15)
The sesquiterpene lactones (STL) helenalin and alantolactone were effective in vitro inhibitors of the mouse hepatic microsomal mixed-function oxidase (MFO) enzymes, aminopyrine demethylase (APD), aniline hydroxylase (ANH) and 7-ethoxyresorufin deethylase (ERD). Helenalin and alantolactone concentrations of 0.5 mM produced a
Xu-Guang Mi et al.
Toxicology letters, 224(3), 349-355 (2013-11-21)
Testes-specific protease 50 (TSP50) is aberrantly expressed in many cancer biopsies and plays a crucial role in tumorigenesis, which make it a potential cancer therapeutic target for drug discovery. Here, we constructed a firefly luciferase reporter driven by the TSP50
J S Ross et al.
Contact dermatitis, 29(2), 84-87 (1993-08-01)
A mix of 3 sesquiterpene lactones (SL) (SL mix 0.1%) was evaluated over a 4-year period. Of 7420 patients with eczema investigated by patch testing, 135 (68 male, 67 female) (1.8%) demonstrated positive reactions, 114 (84%) considered clinically relevant. Females

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service