Skip to Content
Merck
All Photos(2)

Key Documents

A2987

Sigma-Aldrich

Atractylenolide III

≥98% (HPLC)

Synonym(s):

8β-Hydroxyasterolide, Codonolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O3
CAS Number:
Molecular Weight:
248.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

biological source

Atractylodes macrocephala Koidz.

Quality Level

Assay

≥98% (HPLC)

form

powder or crystals

color

white to off-white

solubility

methanol: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

CC1=C2C[C@H]3C(=C)CCC[C@]3(C)C[C@]2(O)OC1=O

InChI

1S/C15H20O3/c1-9-5-4-6-14(3)8-15(17)12(7-11(9)14)10(2)13(16)18-15/h11,17H,1,4-8H2,2-3H3/t11-,14+,15-/m0/s1

InChI key

FBMORZZOJSDNRQ-GLQYFDAESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atractylenolide III is a biologically active sesquiterpene compound, extracted from the roots of Atractylodes macrocephala Koidzumi.

Application

Atractylenolide III has been used to examine its effects on the energy metabolism in the skeletal muscles of mouse.
Atractylenolide III, a sesquiterpenoid, is a phytochemical that may be used to study its anti-inflammatory and anti-angiogenesis activities. Atractylenolide III may be used and studied as an inhibitor of aromatases and inducer of apoptosis. Atractylenolide III may be used to study its sonic hedgehog pathway dependent mechanism of action as an inducer of chondrogenic differentiation. Atractylenolide III may be used as a reference material in assays to detect its presence in plant root extracts and biological milieu such as plasma.

Biochem/physiol Actions

Atractylenolide III has been shown to possess anti-inflammatory effects and also offers gastroprotection. It also shows acaricidal properties and hence has the potential to be used as an agent for the control of dust mites. Atractylenolide III is capable of regulating the secretion and expression of Interleukin-6, thereby mediating the immune response caused by mast cells.
Atractylenolide III possesses anti-inflammatory and anticancer properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun-Teng Wang et al.
The Journal of pharmacy and pharmacology, 62(3), 381-388 (2010-05-22)
The rhizome of Atractylodes ovata De Candolle is popularly used in traditional Chinese medicine to treat gastrointestinal diseases. However, the major gastroprotective compounds of A. ovata have not been identified. This study reports on the principal gastro- protective component of
Chia-Jui Tsai et al.
Bioorganic & medicinal chemistry letters, 22(6), 2326-2329 (2012-03-01)
Two sesquiterpenoids, atractylenolide II and III, were isolated and identified from Atractylodes macrocephala (Asteraceae) to be subsequently evaluated for their activity against farnesoid X receptor (FXR) and progesterone receptor (PR) by transient transfection reporter assays. These sesquiterpenoids did not exert
Hyun-Kyung Kim et al.
Journal of agricultural and food chemistry, 55(15), 6027-6031 (2007-06-28)
The acaricidal activity of materials derived from rhizome of Atractylodes ovata (Atractylodes macrocephala) toward adult Dermatophagoides farinae and Dermatophagoides pteronyssinus was examined using fabric-circle residual contact and vapor-phase toxicity bioassays. Results were compared with those of the currently used acaricides:
Yue Qiu et al.
Evidence-based complementary and alternative medicine : eCAM, 2019, 1392020-1392020 (2020-01-18)
Gastroesophageal reflux disease (GERDs) is a common chronic digestive system disease, in which the symptoms of reflux esophagitis (RE) seriously affect the quality of life. We aimed to study the therapeutic effect of Zhujie Hewei granules (ZHG) on reflux esophagitis
Yuanyuan Fu et al.
Journal of pharmaceutical and biomedical analysis, 174, 595-607 (2019-07-02)
Banxia-Baizhu-Tianma decoction (BBTD) is a compound formulae of traditional Chinese medicine (TCM), which has been clinically used for treatments of neural vertigo, hypertension and epilepsy with a long history. In this study, with an ultra-fast liquid chromatography coupled with quadrupole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service