C1022
Carmine
≥42.0% purity, powder
Synonym(s):
Alum lake of carminic acid, Cochineal, Nacarat, Natural Red 4
About This Item
Recommended Products
Product Name
Carmine, powder, ≥42.0% (UV-vis)
Assay
≥42.0% (UV-vis)
form
powder
color
red
solubility
ammonium hydroxide: 1 mg/mL
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
O1C(C(C(C(C1c2c(c3c(c(c2O)O)C(=O)c4c(c(c(c(c4)O)C(=O)O)C)C3=O)O)O)O)O)CO
InChI
1S/C22H20O13/c1-4-8-5(2-6(24)9(4)22(33)34)13(25)10-11(15(8)27)16(28)12(18(30)17(10)29)21-20(32)19(31)14(26)7(3-23)35-21/h2,7,14,19-21,23-24,26,28-32H,3H2,1H3,(H,33,34)
InChI key
DGQLVPJVXFOQEV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service