Skip to Content
Merck
All Photos(7)

Key Documents

A1653

Sigma-Aldrich

Albumin from human serum

lyophilized powder, ≥96% (agarose gel electrophoresis)

Synonym(s):

HSA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201
NACRES:
NA.25

biological source

human

Assay

≥96% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

monomer calculated mol wt 66478 Da

technique(s)

ELISA: suitable
tissue culture: suitable
western blot: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative

solubility

H2O: soluble 50 mg/mL

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin (ALB) constitutes 25% of the proteins produced by the liver. It is a protein of a single polypeptide chain with a thiol group. The ALB gene is mapped to human 4q13.3.

Application

Albumin from human serum has been used:
  • in cyclophilin isomerase assay
  • as a component in blocking solution for immunogold quantification of amino acids and proteins in complex subcellular compartments
  • in G-pseudotyped HIV (PHIV) assay mixture to study its effect on GSK364735 antiviral potency

Albumin was used to test its effect on the in vitro bactericidal activity of cefditoren against penicillin-resistant Streptococcus pneumonia.
It has been clinically used in serious and often life-threatening conditions, such as shock and blood loss due to trauma, burns, and surgery.

Biochem/physiol Actions

Albumin serves as a protein repository and functions as a binding and transport protein. Albumin binding dissolves the biomolecules in blood and ensures its effective transport. This prevents the loss of substance through excretion. Pathological conditions like vasculitis, lymphangiectasia, glomerulonephritis, and hepatic sinusoidal hypertension might result in hypoalbuminemia.
Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function.

Quality

Remainder mostly globulins

Other Notes

View more information on human serum albumin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluid, electrolyte, and acid-base disturbances in liver disease
Fluid, Electrolyte, and Acid-Base Disorders in Small Animal Practice, 456-456 (2011)
János P Mészáros et al.
Journal of inorganic biochemistry, 195, 91-100 (2019-04-01)
Half-sandwich organometallic complexes of curcumin are extensively investigated as anticancer compounds. Speciation studies were performed to explore the solution stability of curcumin complexes formed with [Rh(η5-C5Me5)(H2O)3]2+. Acetylacetone (Hacac), as the simplest β-diketone ligand bearing (O,O) donor set, was involved for
Angelo Zinellu et al.
European journal of nutrition, 56(2), 785-791 (2015-12-15)
The dietary flavonoids epicatechin (EC), epigallocatechin (EGC), epicatechin gallate (ECG) and epigallocatechin gallate (EGCG) have been shown to interact with circulating albumin for transport in blood to different body tissues. This interaction may modulate their bioavailability and effectiveness. Using affinity
Salvatore Raimondo et al.
Journal of human reproductive sciences, 13(2), 117-124 (2020-08-15)
In the last 10 years, assisted reproductive technologies (ARTs) have offered infertile couples an opportunity to complete their reproductive project. However, the high failure rate could be explained with the complex human reproduction system. In ART, the decrease of the
Serum proteins and the dysproteinemias
Clinical biochemistry of domestic animals, 117-138 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service