Skip to Content
Merck
All Photos(3)

Key Documents

G9885

Sigma-Aldrich

α1-Acid Glycoprotein from human plasma

≥99% (agarose gel electrophoresis)

Synonym(s):

AGP, Orosomucoid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human plasma

Quality Level

Assay

≥99% (agarose gel electrophoresis)

form

powder

technique(s)

ELISA: suitable

impurities

HIV-1/HIV-2, HCV and hepatitis B antigen, tested negative

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... ORM1(5004)

Looking for similar products? Visit Product Comparison Guide

General description

α-1-Acid glycoprotein (AGP)/orosomucoid (ORM), a glycoprotein has a molecular weight of 41-43kDa. It is considered as one of the important acute phase proteins in humans and rats. AGP may belong to the lipocalin family.[1]

Application

α1-Acid glycoprotein from human plasma has been used:
  • as negatively charged protein to investigate the adsorption processes onto the surface of polyelectrolyte multilayers[2]
  • to incubate plates coated with purified antiserum in the study to investigate its serum concentration by ELISA[3]
  • to study the effect of its different concentrations on the antiviral activity of human immunodeficiency virus (HIV) protease inhibitors[4]

Biochem/physiol Actions

α1-Acid glycoprotein possesses immunomodulating effects. It has the ability to bind to acidic drugs, like phenobarbital.[1]

Preparation Note

Purified from Cohn Fraction VI

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chenhua Zhang et al.
Analytica chimica acta, 1078, 189-199 (2019-07-31)
Silica-based lectin microcolumns were developed and optimized for the separation and analysis of glycoform fractions in alpha1-acid glycoprotein (AGP) based on both the degree of branching and level of fucosylation. Concanavalin A (Con A) and Aleuria Aurantia lectin (AAL) were
In Vitro Effect of alpha1-Acid Glycoprotein on the Anti?Human Immunodeficiency Virus (HIV) Activity of the Protease Inhibitor CGP 61755: A Comparative Study with Other Relevant HIV Protease Inhibitors
Lazdins J K, et al.
The Journal of Infectious Diseases, 175(5), 1063-1070 (1997)
Hendrik Grootaert et al.
Glycobiology, 30(9), 735-745 (2020-03-10)
The deoxy sugar l-fucose is frequently found as a glycan constituent on and outside living cells, and in mammals it is involved in a wide range of biological processes including leukocyte trafficking, histo-blood group antigenicity and antibody effector functions. The
Effect of once weekly administration of mifepristone on ovarian function in normal women
Chen X and Xiao B
Contraception, 56(3), 175-180 (1997)
Protein adsorption onto auto-assembled polyelectrolyte films
Ladam G, et al.
Langmuir, 17(3), 878-882 (2001)

Questions

  1. Please, could you tell me what are the best storage conditions once reconstituted in water, and for how long are reconstituted solutions stable?

    1 answer
    1. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for to 3 months at (-20°C).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service