Skip to Content
Merck
All Photos(6)

Key Documents

A3782

Sigma-Aldrich

Albumin from human serum

lyophilized powder, Fatty acid free, Globulin free, ≥99% (agarose gel electrophoresis)

Synonym(s):

HSA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201
NACRES:
NA.25

biological source

human

Assay

≥99% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

monomer calculated mol wt 66478 Da

technique(s)

ELISA: suitable
tissue culture: suitable
western blot: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative
≤0.1% Fatty acids

solubility

H2O: soluble 50 mg/mL

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin is the most abundant protein in blood plasma, produced in the liver.

Albumin is a known carrier of fatty acids (FA). Thus control over specific FA′s for cell culture is important, as different cell lines can differ in their sensitivity to particular fatty acids. Fatty acid-free human serum albumin (HSA) is therefore useful for cell culture studies where specific fatty acid content must be strictly controlled, so that researchers can use particular fatty acids specific to their cell lines. Fatty acid-free albumin also allows for optimal and maximum binding sites for using specific fatty acids in cell culture. The use of FA-free HSA also addresses concerns about endogenous FA′s potentially in non-FA-free HSA.
Albumin is the most copious plasma protein in humans. Albumin turnover is seen in infants with iron deficiency anemia. Serum albumin is a reliable prognostic indicator in liver disease. Oxidative damage of albumin is associated with advanced liver disease.

Albumin is widely used as a blocking agent in immunoassays and immunodetection procedures, and as a carrier protein for dilution of antibodies. Awareness of potential trace endogenous content of globulins / IgG in human serum albumin (HSA) is therefore important, because such trace globulins / IgG may serve as antigens for secondary antibodies in immunodetection work. This product undergoes testing for low globulin content / status as essentially globulin-free.

Application

Albumin from human serum has been used in the preparation of glycated human serum albumin (gHSA) in a glucose solution. It has also been used in the study to estimate the impact of surface nanotopography and chemical composition on blood compatibility.
Albumin was used to test its effect on the in vitro bactericidal activity of cefditoren against penicillin-resistant Streptococcus pneumonia. It has been clinically used in serious and often life-threatening conditions, such as shock and blood loss due to trauma, burns, and surgery. It was used also to test the effect of non-enzymatic glycation on the unfolding of human serum albumin.

Biochem/physiol Actions

Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function.

Features and Benefits

  • Fatty acid-free / low fatty acid content
  • Globulin-free / low globulin content

Other Notes

View more information on human serum albumin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudio Dalvit et al.
Magnetic resonance in chemistry : MRC, 55(2), 106-114 (2016-08-16)
Ligand-based
Competitive adsorption of fibrinogen and albumin and blood platelet adhesion on surfaces modified with nanoparticles and/or PEO
Nonckreman C J, et al.
Colloids and Surfaces. B, Biointerfaces, 77(2), 139-149 (2010)
Fibrillation and Polymorphism of Human Serum Albumin
Bio-nanoimaging, 77(2), 345-362 (2014)
Isabel Correia et al.
Chemistry, an Asian journal, 12(16), 2062-2084 (2017-06-27)
[VO(acac)
Coordination ability and biological activity of a naringenin thiosemicarbazone.
Katarzyna Brodowska et al.
Journal of inorganic biochemistry, 165, 36-48 (2016-10-21)

Articles

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service