Skip to Content

A9080

Albumin solution human

30% in 0.85% sodium chloride, protease free

Synonym(s):

HSA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25
MDL number:
eCl@ss:
42010201
Biological source:
human
Form:
liquid
Technique(s):
ELISA: suitable, tissue culture: suitable, western blot: suitable
Impurities:
HIV I and HIVII, HCV and HBsAg, tested negative
Concentration:
29.5-35.0% protein (biuret), 30% in 0.85% sodium chloride
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

form

liquid

concentration

29.5-35.0% protein (biuret), 30% in 0.85% sodium chloride

technique(s)

ELISA: suitable, tissue culture: suitable, western blot: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

General description

Albumin is the most copious protein in blood plasma. Liver produces human albumin. Human serum albumin undergoes three different post-translational modifications: oxidation, glycation, and S-nitrosylation. Modifications usually occur on the surface of the globular protein, and do not significantly affect conformation. However, modification strongly affects binding of fatty acids and drug molecules.
Human serum albumin undergoes three different post-translational modifications: oxidation, glycation, and S-nitrosylation. Modifications usually occur on the surface of the globular protein, and do not significantly affect conformation. However, modification strongly affects binding of fatty acids and drug molecules.

Other Notes

View more information on human serum albumin.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.


Still not finding the right product?

Explore all of our products under Albumin solution human


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library