Skip to Content
Merck
All Photos(1)

Key Documents

94378

Sigma-Aldrich

Methyl cellulose

viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)

Synonym(s):

Methyl cellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.25

form

powder

Quality Level

viscosity

3000-5500 mPa.s, 2 % in H2O(20 °C)

SMILES string

[*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]

InChI key

YLGXILFCIXHCMC-JHGZEJCSSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Methyl cellulose is a gum polysaccharide containing glucose with methyl substitution.

Application

Methyl cellulose has been used:


  • in polymer solution to enhance viscosity in the formulation for dip coating
  • in growth medium to facilitate stable morphology
  • as a supplement in HMI-9 medium to increase viscosity

Biochem/physiol Actions

Methyl cellulose is known to have a molecular size that might pass through placenta. It is useful in visualizing small bowel loops through its overlaps.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bronchopulmonary infection due to Branhamella catarrhalis.
DT McLeod, F Ahmad, et al.
British Medical Journal, 287, 1446-1447 (1983)
R Olsson et al.
Acta radiologica (Stockholm, Sweden : 1987), 38(1), 73-75 (1997-01-01)
Double-contrast barium enema has a reduced sensitivity in patients with severe sigmoid diverticulosis. Therefore a carboxy methyl cellulose enema was employed after the conventional double-contrast examination in 15 patients with sigmoid diverticulosis. A significant increase in lumen diameter and a
Drugs for Pregnant and Lactating Women E-Book (2009)
Sarah Schuster et al.
eLife, 10 (2021-08-07)
African trypanosomes cause sleeping sickness in humans and nagana in cattle. These unicellular parasites are transmitted by the bloodsucking tsetse fly. In the mammalian host's circulation, proliferating slender stage cells differentiate into cell cycle-arrested stumpy stage cells when they reach
Abdominal Imaging E-Book: Expert Radiology Series (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service