Skip to Content
Merck
All Photos(3)

Key Documents

423238

Sigma-Aldrich

(Hydroxypropyl)methyl cellulose

average Mn ~10,000

Synonym(s):

HPMC, Hypromellose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

average Mn ~10,000

extent of labeling

1.8-2.0 mol methoxy per mol cellulose (D.S.)
29 wt. % methoxy
0.2-0.3 mol propylene oxide per mol cellulose (M.S.)
7 wt. % propylene oxide

surface tension

43-55 dyn/cm, 25 °C, 0.05 wt. %

viscosity

6 cP(20 °C)(lit.)

transition temp

gel point 58-64 °C

SMILES string

O3[C@H]([C@@H]([C@H]([C@@H]([C@H]3COCC(O)C)OCC(O)C)OCC(O)C)OCC(O)C)O[C@@H]4[C@H](O[C@H]([C@@H]([C@H]4OCC(O)C)OCC(O)C)OCC(O)C)COCC(O)C.O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1COC)OC)OC)OC)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2OC)OC)OC)COC

InChI key

PUSNGFYSTWMJSK-GSZQVNRLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thickener for aqueous and non-aqueous systems, clear films with grease resistance, binders, lubricants, steric stabilizer and water retention aid.

Features and Benefits

Dissolves in water, undergoes reversible gelation upon heating, non-ionic, does not complex with ionic species and is surface active and enzyme resistant. Solutions are pseudoplastic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeonghoon Han et al.
Aquatic toxicology (Amsterdam, Netherlands), 152, 308-317 (2014-05-13)
In this study, we investigated the effects of the water-accommodated fraction (WAF) of crude oil on the development and reproduction of the intertidal copepod Tigriopus japonicus through life-cycle experiments. Furthermore, we investigated the mechanisms underlying the toxic effects of WAF
Fernando Andrés et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(26), E2760-E2769 (2014-07-01)
In Arabidopsis thaliana environmental and endogenous cues promote flowering by activating expression of a small number of integrator genes. The MADS box transcription factor SHORT VEGETATIVE PHASE (SVP) is a critical inhibitor of flowering that directly represses transcription of these
Somayeh Bahrami et al.
Veterinary parasitology, 203(1-2), 43-50 (2014-03-19)
Fasciola gigantica is a parasitic helminth that predominantly infects the liver and bile ducts of cattle and causes great losses of cattle production in the southwestern regions of Iran. The purpose of the present study was to find out the
Jane W Agger et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(17), 6287-6292 (2014-04-16)
The recently discovered lytic polysaccharide monooxygenases (LPMOs) are known to carry out oxidative cleavage of glycoside bonds in chitin and cellulose, thus boosting the activity of well-known hydrolytic depolymerizing enzymes. Because biomass-degrading microorganisms tend to produce a plethora of LPMOs
Youcai Hu et al.
Nature chemical biology, 10(7), 552-554 (2014-05-20)
Despite the remarkable versatility displayed by flavin-dependent monooxygenases (FMOs) in natural product biosynthesis, one notably missing activity is the oxidative generation of carbonate functional groups. We describe a multifunctional Baeyer-Villiger monooxygenase, CcsB, which catalyzes the formation of an in-line carbonate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service