Skip to Content
Merck
All Photos(1)

Key Documents

Y0001460

Acetylsalicylic acid for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Acetylsalicylic acid, 2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
779271
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

aspirin

manufacturer/tradename

EDQM

mp

134-136 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccccc1C(O)=O

InChI

1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)

InChI key

BSYNRYMUTXBXSQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acetylsalicylic acid for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Blocks the production of prostaglandins by inhibiting cyclooxygenase (prostaglandin H synthase), with greater selectivity toward the COX-1 isoform. The antithrombotic effect is due to the inhibition of COX-1 in platelets that blocks thromboxane production and platelet aggregation. Chemopreventive against colorectal and other solid tumors.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

482.0 °F

Flash Point(C)

250 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurent Arnaud et al.
Autoimmunity reviews, 13(3), 281-291 (2013-11-06)
We performed a meta-analysis to determine whether aspirin has a significant protective effect on risk of first thrombosis among patients with antiphospholipid antibodies (aPL+). Observational and interventional studies identified from the Medline, Embase and Cochrane databases were selected if they
P J Devereaux et al.
The New England journal of medicine, 370(16), 1494-1503 (2014-04-01)
There is substantial variability in the perioperative administration of aspirin in patients undergoing noncardiac surgery, both among patients who are already on an aspirin regimen and among those who are not. Using a 2-by-2 factorial trial design, we randomly assigned
Ahmed Kayssi et al.
Journal of vascular surgery, 60(1), 106-110 (2014-03-25)
Acute limb ischemia (ALI) in pediatric patients is rare but may lead to limb loss and life-long complications. This study reviewed the experience of a Canadian tertiary pediatric center with the medical and operative management of ALI. The medical records of
Naomi Gronich et al.
Nature reviews. Clinical oncology, 10(11), 625-642 (2013-10-02)
Cancer risk reduction using pharmacological means is an attractive modern preventive approach that supplements the classical behavioural prevention recommendations. Medications that are commonly used by large populations to treat a variety of common, non-cancer-related, medical situations are an attractive candidate
Rickard Ljung et al.
International journal of cancer, 135(1), 238-241 (2013-12-19)
Anticoagulant treatment might enhance the natural defense against tumor cell dissemination caused by diagnostic needle biopsy by counteracting thrombocyte coating of such cells. To clarify whether women using anticoagulant treatment at the time of biopsy have a lower occurrence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service