Skip to Content
Merck

Skip To

A5376

Acetylsalicylic acid

≥99.0%

Synonym(s):

2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
UNSPSC Code:
12352106
NACRES:
NA.21
PubChem Substance ID:
EC Number:
200-064-1
Beilstein/REAXYS Number:
779271
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Level

assay

≥99.0%

form

powder

color

white

pKa (25 °C)

3.5

mp

134-136 °C (lit.)

solubility

H2O: 10 mg/mL at 37 °C(lit.), H2O: 3 mg/mL at 25 °C(lit.), ethanol: 50 mg/mL, aqueous base: decomposes

ε (extinction coefficient)

190 at 298 nm in aqueous base at 1 mM, 409 at 231 nm in aqueous base at 1 mM, 466 at 230 nm in aqueous acid at 1 mM, 68 at 278 nm in aqueous acid at 1 mM

SMILES string

CC(=O)Oc1ccccc1C(O)=O

InChI

1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)

InChI key

BSYNRYMUTXBXSQ-UHFFFAOYSA-N

Gene Information

General description

Acetylsalicylic acid is an organic compound commonly referred to as aspirin. It works by blocking the production of compounds in the body that cause pain, swelling, and fever. Acetylsalicylic acid also serves a vital function in preventing blood clots from forming in the arteries. In research, acetylsalicylic acid is studied for its role in oxidative stress, and in cancer research as a biomarker.

Biochem/physiol Actions

Blocks the production of prostaglandins by inhibiting cyclooxygenase (prostaglandin H synthase), with greater selectivity toward the COX-1 isoform. The antithrombotic effect is due to the inhibition of COX-1 in platelets that blocks thromboxane production and platelet aggregation. Chemopreventive against colorectal and other solid tumors.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A0200000A2093BP617
Aspirin British Pharmacopoeia (BP) Reference Standard

BP617

Aspirin

assay

≥99.0%

assay

-

assay

-

assay

-

biological source

synthetic

biological source

-

biological source

-

biological source

-

Gene Information

human ... ITGA2B(3674), ITGB3(3690), PTGS1(5742), PTGS2(5743)

Gene Information

human ... PTGS1(5742), PTGS2(5743)

Gene Information

human ... ITGA2B(3674), PTGS1(5742), PTGS2(5743)

Gene Information

human ... PTGS1(5742), PTGS2(5743)

Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

-

form

powder

form

-

form

powder or crystals

form

crystalline

solubility

H2O: 10 mg/mL at 37 °C(lit.), ethanol: 50 mg/mL, H2O: 3 mg/mL at 25 °C(lit.), aqueous base: decomposes

solubility

-

solubility

-

solubility

-


Still not finding the right product?

Explore all of our products under Acetylsalicylic acid


pictograms

Exclamation mark

signalword

Warning

Storage Class

11 - Combustible Solids

flash_point_f

482.0 °F

flash_point_c

250 °C

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Seung-Hwan Jung et al.
Redox biology, 37, 101751-101751 (2020-10-21)
Treatment with nonsteroidal anti-inflammatory drugs (NSAIDs) is associated with various side effects, including cardiovascular and hepatic disorders. Studies suggest that mitochondrial damage and oxidative stress are important mediators of toxicity, yet the underlying mechanisms are poorly understood. In this study
Niels Hulsman et al.
Journal of medicinal chemistry, 50(10), 2424-2431 (2007-04-20)
Hybrid drug 1 (NO-ASA) continues to attract intense research from chemists and biologists alike. It consists of ASA and a -ONO2 group connected through a spacer and is in preclinical development as an antitumor drug. We report that, contrary to
Connie N Hess et al.
Journal of the American College of Cardiology, 66(7), 777-787 (2015-08-14)
Bleeding limits anticoagulant treatment in patients with acute coronary syndromes (ACS). We investigated whether background concomitant antiplatelet therapy influences the effects of apixaban after ACS. This study examined high-risk ACS patients who were treated with aspirin or aspirin plus clopidogrel



Global Trade Item Number

SKUGTIN
A5376-250G04061833375815
A5376-100G04061835542253
A5376-500G04061835542260
A5376-1KG04061833375808

Questions

1–8 of 8 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. This product, A5376, could be used for cell culture ? What about the stability of the product ?

    1 answer
    1. In the original sealed container, this material is quited stable and assigned a retest date of 5 years from the date of Quality Release. The retest date is lot specific and published in the Certificate of Analysis. In aqueous solution, Acetylsalicylic acid is hydrolyzed gradually to salicylic acid. Please see the link below to review a publication ragarding the half-life in solution:
      https://www.sciencedirect.com/science/article/pii/S0095955315360510

      This product has not been tested for suitability in cell culture. However, researchers have cited using this product for cell culture applications. Please see the links below to review published examples:
      https://www.nature.com/articles/s41467-017-00259-3
      https://www.sciencedirect.com/science/article/pii/S0887233302001285

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. could you please tell me what is the stability of aspirin (cat #5376) once recnstituted in PBS please?

    1 answer
    1. Aqueous solutions of acetylsalicylic acid will readily hydrolyze to form salicylic acid. Solutions are most stable at pH 2 - 3, but to assure full activity, solutions should be freshly prepared. Please see the link below to review a publication on the compound stability:
      https://intapi.sciendo.com/pdf/10.1515/cipms-2016-0009

      Helpful?

  5. How does aspirin act as an anticlotting agent?

    1 answer
    1. There may be multiple modes of action which result in the anticlotting properties of aspirin. Some studies have indicated a decrease in thrombin levels and others reduced clotting properties of fibrinogen.

      Helpful?

  6. Is Product A5376, Acetylsalicylic acid, offered by Sigma-Aldrich different from that found in aspirin tablets from the drug store?

    1 answer
    1. The active ingredient acetylsalicylic acid offered by Sigma-Aldrich is the same chemical as found in aspirin tablets. However, chemicals for pharmaceutical use are manufactured, tested, and packaged under the regulation of the FDA, but chemicals sold for research use do not have the same regulatory oversight.

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  8. How should solutions of Product A5376, Acetylsalicylic acid, be prepared and stored?

    1 answer
    1. Acetylsalicylic acid is slightly soluble in water, with a limit of solubility reported as approximately 3 mg/ml at 25 Deg C.  It is also soluble in ethanol at 50 mg/ml and will dissolve in solutions of alkali hydroxides and carbonates with decomposition.  Aqueous solutions of acetylsalicylic acid are most stable at pH 2 - 3, but to assure full activity, fresh solutions are recommended.

      Helpful?

Reviews

No rating value

Active Filters