Skip to Content
Merck
All Photos(1)

Key Documents

Y0000849

Clotrimazole for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Clotrimazole, 1-(o-Chloro-α,α-diphenylbenzyl)imidazole, 1-(o-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17ClN2
CAS Number:
Molecular Weight:
344.84
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clotrimazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1ccccc1C(c2ccccc2)(c3ccccc3)n4ccnc4

InChI

1S/C22H17ClN2/c23-21-14-8-7-13-20(21)22(25-16-15-24-17-25,18-9-3-1-4-10-18)19-11-5-2-6-12-19/h1-17H

InChI key

VNFPBHJOKIVQEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clotrimazole for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria F Sassano et al.
Journal of medicinal chemistry, 56(6), 2406-2414 (2013-02-27)
Colloidal aggregation is the dominant mechanism for artifactual inhibition of soluble proteins, and controls against it are now widely deployed. Conversely, investigating this mechanism for membrane-bound receptors has proven difficult. Here we investigate the activity of four well-characterized aggregators against
Jenna Corcoran et al.
Aquatic toxicology (Amsterdam, Netherlands), 152, 57-65 (2014-04-15)
Clotrimazole (CTZ) is a persistent imidazole antifungal agent which is frequently detected in the aquatic environment and predicted to bio-concentrate in fish. Common carp (Cyprinus carpio) were exposed to mean measured concentrations of either 1.02 or 14.63μgl(-1) CTZ for 4
Laura Ravani et al.
International journal of pharmaceutics, 454(2), 695-702 (2013-06-25)
The aim of the present work is to design a new formulation containing clotrimazole (CTZ) loaded into nanostructured lipid carriers (NLC) for the treatment of fungal vaginal infections. In order to obtain formulations with suitable viscosity for mucosal application, NLC
Sandra Gemma et al.
Journal of medicinal chemistry, 55(15), 6948-6967 (2012-07-13)
Despite recent progress in the fight against malaria, the emergence and spread of drug-resistant parasites remains a serious obstacle to the treatment of infections. We recently reported the development of a novel antimalarial drug that combines the 4-aminoquinoline pharmacophore of
Yingzi Zhao et al.
The Journal of pharmacology and experimental therapeutics, 344(1), 15-22 (2012-09-26)
In the aorta of male spontaneously hypertensive rats (SHR), but not in that of normotensive Wistar-Kyoto rats (WKY), contractions to phenylephrine obtained in the presence of L-NAME [inhibitor of nitric oxide synthase (NOS)] and indomethacin (inhibitor of cyclooxygenase) are inhibited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service