Skip to Content
Merck
All Photos(1)

Key Documents

C2430000

Clotrimazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(o-Chloro-α,α-diphenylbenzyl)imidazole, 1-(o-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H17ClN2
CAS Number:
Molecular Weight:
344.84
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clotrimazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Clc1ccccc1C(c2ccccc2)(c3ccccc3)n4ccnc4

InChI

1S/C22H17ClN2/c23-21-14-8-7-13-20(21)22(25-16-15-24-17-25,18-9-3-1-4-10-18)19-11-5-2-6-12-19/h1-17H

InChI key

VNFPBHJOKIVQEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clotrimazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Natalia Mast et al.
Molecular pharmacology, 84(1), 86-94 (2013-04-23)
Although there are currently three generations of antifungal azoles on the market, even the third-generation agents show undesirable interactions with human cytochrome P450 (P450) enzymes. CYP46A1 is a cholesterol-metabolizing P450 in the brain that tightly binds a number of structurally
Catherine L Neary et al.
Journal of cellular physiology, 228(9), 1943-1948 (2013-05-01)
Hexokinase II (HK2), the enzyme that catalyzes the first committed step of glycolysis, is overexpressed in many cancers, as is the central signaling kinase Akt. Akt activity promotes HK2 association with the mitochondria, as well as glucose uptake by cancer
Yingzi Zhao et al.
The Journal of pharmacology and experimental therapeutics, 344(1), 15-22 (2012-09-26)
In the aorta of male spontaneously hypertensive rats (SHR), but not in that of normotensive Wistar-Kyoto rats (WKY), contractions to phenylephrine obtained in the presence of L-NAME [inhibitor of nitric oxide synthase (NOS)] and indomethacin (inhibitor of cyclooxygenase) are inhibited
Sandra Gemma et al.
Journal of medicinal chemistry, 55(15), 6948-6967 (2012-07-13)
Despite recent progress in the fight against malaria, the emergence and spread of drug-resistant parasites remains a serious obstacle to the treatment of infections. We recently reported the development of a novel antimalarial drug that combines the 4-aminoquinoline pharmacophore of
Laura Ravani et al.
International journal of pharmaceutics, 454(2), 695-702 (2013-06-25)
The aim of the present work is to design a new formulation containing clotrimazole (CTZ) loaded into nanostructured lipid carriers (NLC) for the treatment of fungal vaginal infections. In order to obtain formulations with suitable viscosity for mucosal application, NLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service