Skip to Content
Merck

Skip To

P9623

Paroxetine hydrochloride hemihydrate

≥98% (HPLC), powder, serotonin reuptake inhibitor

Synonym(s):

Paroxetine HCl, (3S-trans)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride hemihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H20FNO3 · HCl · .5 H2O
CAS Number:
Molecular Weight:
374.83
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Paroxetine hydrochloride hemihydrate, ≥98% (HPLC), powder

Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

originator

GlaxoSmithKline

SMILES string

O.Cl[H].Fc1ccc(cc1)[C@@H]2CCNC[C@H]2COc3ccc4OCOc4c3

InChI

1S/C19H20FNO3.ClH.H2O/c20-15-3-1-13(2-4-15)17-7-8-21-10-14(17)11-22-16-5-6-18-19(9-16)24-12-23-18;;/h1-6,9,14,17,21H,7-8,10-12H2;1H;1H2/t14-,17-;;/m0../s1

InChI key

QRQSGFFISBKLMZ-YHOFXEKLSA-N

Gene Information

human ... SLC6A4(6532)

Application

Paroxetine hydrochloride hemihydrate has been used: as an antidepressant to study its effects on neural stem cells (NSCs) from embryonic rat hippocampus in vitro[1]; as a CYP 2D6 inhibitor to study its effects on the in vitro production by human recombinant CYP 2D6 of the phenolic metabolites; as an antidepressant to study its effects on the expression of pituitary adenylate cyclase activating polypeptide (PACAP), its receptors and brain-derived neurotrophic factor (BDNF) in rat primary hippocampal neurons

Biochem/physiol Actions

Paroxetine hydrochloride hemihydrate is one of the most potent and selective of the selective serotonin reuptake inhibitors (SSRI); antidepressant
Selective serotonin reuptake inhibitor; antidepressant
Paroxetine is a phenylpiperidine derivative. It has the ability to cross placenta. Paroxetine is known to increase the risk of congenital cardiac malformations. It might be useful in hormone replacement therapy for treating vasomotor symptoms during menopause. It is known to ameliorate the effects of panic disorder, obsessive‐compulsive disorder and social phobia.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold with the permission of GlaxoSmithKline

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP1024Y0000281PHR1804
assay

≥98% (HPLC)

assay

-

assay

-

assay

-

form

powder

form

powder

form

-

form

-

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

300

storage condition

desiccated

storage condition

-

storage condition

-

storage condition

-

color

white

color

-

color

-

color

-

originator

GlaxoSmithKline

originator

-

originator

-

originator

-


Still not finding the right product?


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters