Skip to Content
Merck
All Photos(2)

Key Documents

F124

Sigma-Aldrich

Furafylline

≥98% (HPLC), powder, caffeine inhibitor

Synonym(s):

3-(2-Furanylmethyl)-3,7-dihydro-1,8-dimethyl-1H-purine-2,6-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N4O3
CAS Number:
Molecular Weight:
260.25
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Product Name

Furafylline, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

mp

274-275  °C

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

CN1C(=O)N(Cc2ccco2)c3nc(C)[nH]c3C1=O

InChI

1S/C12H12N4O3/c1-7-13-9-10(14-7)16(6-8-4-3-5-19-8)12(18)15(2)11(9)17/h3-5H,6H2,1-2H3,(H,13,14)

InChI key

KGQZGCIVHYLPBH-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Biochem/physiol Actions

Furafylline (1,8-dimethyl-3-(2′ -furfuryl)methylxanthine) is a xanthine derivative. It is preferred in treating asthma. It serves as a N3-demethylation inhibitor of caffeine. Furafylline does not show much effect on human monooxygenase activities. It is considered as an efficient bronchodilator and as an inhibitor of anaphylactic reactions, when compared to theophylline.
Furafylline is a methyl xanthine derivative with longer duration of action than theophylline and an inhibitor of cytochrome P4501A2.

Features and Benefits

This compound is a featured product for ADME Tox and Cyclic Nucleotide research. Discover more featured ADME Tox and Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Labbé et al.
Xenobiotica; the fate of foreign compounds in biological systems, 33(1), 13-25 (2003-01-10)
1. Mexiletine is extensively metabolized in man by C- and N-oxidation and the aim of the present study was to characterize major cytochrome P450 enzyme(s) involved in the formation of N-hydroxymexiletine. 2. Incubations with genetically engineered microsomes indicated that the
Marilia N Siebert et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 203, 115-121 (2016-10-23)
Cytochrome P450 family 1 (CYP1) is involved in polycyclic aromatic hydrocarbons (PAHs) biotransformation. PAHs can induce CYP1 protein expression and enzyme activity, the latter being usually quantified as 7-ethoxyresorufin O-deethylase activity (EROD). The aim of this study was to characterize
Nicholas Ekow Thomford et al.
Journal of ethnopharmacology, 211, 58-69 (2017-09-25)
Hyptis suaveolens (L) Poit and Boerhavia diffusa Linn are medicinal herbal plants commonly found in the tropics and sub-tropics. They are used to treat various conditions among them boils, dyslipidaemia, eczema, malaria, jaundice and gonorrhoea. Thus, the herbal medicinal extracts
Accumulation of caffeine in healthy volunteers treated with furafylline.
Tarrus E, et al.
British Journal of Clinical Pharmacology, 23(1), 9-18 (1987)
Elisa Ollikainen et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 104, 124-132 (2017-04-04)
Engineered nanoparticles are increasingly used as drug carriers in pharmaceutical formulations. This study focuses on the hitherto unaddressed impact of porous silicon (PSi) nanoparticles on human cytochrome P450 (CYP) metabolism, which is the major detoxification route of most pharmaceuticals and

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service