Skip to Content
Merck

Skip To

K1003

Ketoconazole

99.0-101.0% (EP, titration), meets EP testing specifications

Synonym(s):

(±)-cis-1-Acetyl-4-(4-[(2-[2,4-dichlorophenyl]-2-[1H-imidazol-1-ylmethyl]-1,3-dioxolan-4-yl)-methoxy]phenyl)piperazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C26H28Cl2N4O4
CAS Number:
Molecular Weight:
531.43
NACRES:
NA.85
PubChem Substance ID:
UNSPSC Code:
51101500
EC Number:
265-667-4
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

meets EP testing specifications

description

Specific Optical Rotation (EP): (−0.10) ∼ +0.10 °

assay

99.0-101.0% (EP, titration)

form

powder

color

white to off-white

antibiotic activity spectrum

Gram-positive bacteria, fungi, yeast

mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

CC(=O)N1CCN(CC1)c2ccc(OC[C@H]3CO[C@@](Cn4ccnc4)(O3)c5ccc(Cl)cc5Cl)cc2

InChI

1S/C26H28Cl2N4O4/c1-19(33)31-10-12-32(13-11-31)21-3-5-22(6-4-21)34-15-23-16-35-26(36-23,17-30-9-8-29-18-30)24-7-2-20(27)14-25(24)28/h2-9,14,18,23H,10-13,15-17H2,1H3/t23-,26-/m0/s1

InChI key

XMAYWYJOQHXEEK-OZXSUGGESA-N

General description

Chemical structure: imidazole

Application

Ketoconazole is a broad spectrum antifungal agent used to treat candidiasis, chronic mucocutaneous candidiasis, oral thrush, candiduria, blastomycosis, coccidioidomycosis, histoplasmosis, chromomycosis, and paracoccidioidomycosis. It is used to identify p-glycoprotein/CYP3A-limited bioavailability in the monkey model, to study interleukin 1 mediated antitumor effects, and drug interactions in vivo
CYP3A4 inhibitor

Biochem/physiol Actions

Antifungal agent
Ketoconazole interacts with 14-α demethylase, a cytochrome P-450 enzyme that is necessary for the conversion of lanosterol to ergosterol. This interaction inhibits ergosterol synthesis and results in increased fungal cellular permeability. Other possible mechanisms of action are the inhibition of endogenous respiration, interaction with membrane phospholipids, inhibition of yeast transformation to mycelial forms, inhibition of purine uptake, and impairment of triglyceride and/or phospholipid biosynthesis. Ketoconazole inhibits the synthesis of thromboxane and sterols such as aldosterone, cortisol, and testosterone .

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1356508BP1170BP1169
description

99.0-101.0% (EP, titration), meets EP testing specifications

description

United States Pharmacopeia (USP) Reference Standard

description

British Pharmacopoeia (BP) Reference Standard

description

British Pharmacopoeia (BP) Reference Standard

mode of action

enzyme | inhibits

mode of action

-

mode of action

-

mode of action

-

antibiotic activity spectrum

fungi, yeast, Gram-positive bacteria

antibiotic activity spectrum

-

antibiotic activity spectrum

-

antibiotic activity spectrum

-

Quality Level

200

Quality Level

-

Quality Level

-

Quality Level

-

assay

99.0-101.0% (EP, titration)

assay

-

assay

-

assay

-

form

powder

form

-

form

powder

form

powder

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Ketoconazole


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters