Skip to Content
Merck

Skip To

T6376

Triamcinolone

Glucocorticoid agonist

Synonym(s):

9α-Fluoro-11β,16α,17,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-11β,16α,17,21-tetrahydroxypregna-1,4-diene-3,20-dione, Fluoxyprednisolone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H27FO6
CAS Number:
Molecular Weight:
394.43
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
204-718-7
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Triamcinolone,

Quality Segment

mp

262-263 °C (lit.)

solubility

DMF: soluble 20 mg/mL, methanol: soluble 5 mg/mL, DMSO: soluble (Refer citation 16)

originator

Abbott

SMILES string

[H][C@@]12C[C@@H](O)[C@](O)(C(=O)CO)[C@@]1(C)C[C@H](O)[C@@]3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C21H27FO6/c1-18-6-5-12(24)7-11(18)3-4-13-14-8-15(25)21(28,17(27)10-23)19(14,2)9-16(26)20(13,18)22/h5-7,13-16,23,25-26,28H,3-4,8-10H2,1-2H3/t13-,14-,15+,16-,18-,19-,20-,21-/m0/s1

InChI key

GFNANZIMVAIWHM-OBYCQNJPSA-N

Gene Information

Application

Triamcinolone has been used as a corticosteroid standard for LC-MS-MS analysis. Triamcinolone has also been used to study its effect on IgG and IgE levels in peripheral blood mononuclear cells (PBMCs)[1]

Biochem/physiol Actions

Triamcinolone is a synthetic glucocorticoid agonist; induces gene expression and apoptosis; inhibits prostaglandin synthesis; impairs tumor necrosis factor (TNF)-α-induced degradation of κB-α; potentiates the differentiation-inducing effects of bone morphogenetic proteins (BMP-2, -4, -6).

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T6501T18999901676000
Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

-

solubility

DMF: soluble 20 mg/mL, DMSO: soluble (Refer citation 16), methanol: soluble 5 mg/mL

solubility

-

solubility

-

solubility

-

Gene Information

human ... NR3C1(2908), PTGS2(5743)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

mp

262-263 °C (lit.)

mp

-

mp

262-263 °C (lit.)

mp

262-263 °C (lit.)

originator

Abbott

originator

-

originator

-

originator

-


Still not finding the right product?

Explore all of our products under Triamcinolone


pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters