Skip to Content
Merck
All Photos(1)

Key Documents

46074

Supelco

Betamethasone 17-valerate

VETRANAL®, analytical standard

Synonym(s):

1,4-Pregnadiene-11β,17α,21-triol-9α-fluoro-16β-methyl-3,20-dione 17-valerate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17-valerate, 9α-Fluoro-16β-methylprednisolone 17-valerate, Betnovate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37FO6
CAS Number:
Molecular Weight:
476.58
Beilstein:
4240001
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical

format

neat

storage temp.

2-8°C

SMILES string

CCCCC(=O)O[C@@]1([C@@H](C)CC2C3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO

InChI

1S/C27H37FO6/c1-5-6-7-23(33)34-27(22(32)15-29)16(2)12-20-19-9-8-17-13-18(30)10-11-24(17,3)26(19,28)21(31)14-25(20,27)4/h10-11,13,16,19-21,29,31H,5-9,12,14-15H2,1-4H3/t16-,19?,20?,21-,24-,25-,26-,27-/m0/s1

InChI key

SNHRLVCMMWUAJD-QDHNOTTGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Saif-ur-Rehman Khattak et al.
AAPS PharmSciTech, 14(1), 177-182 (2012-12-20)
The effects of solvent [acetonitrile, methanol, and acetonitrile/water mixture (20:80, v/v)], buffer concentration (phosphate buffer, pH 7.5), ionic strength and commonly employed adjuvants on the photodegradation of betamethasone-17 valerate in cream and gel formulations have been studied on exposure to UV
Jens-Michael Jensen et al.
Experimental dermatology, 20(10), 783-788 (2011-06-29)
It has been suggested that the increased rate of bacterial infection in atopic dermatitis (AD) may be caused by reduced antimicrobial protein (AMP) expression. We were interested whether common treatments in AD affect antimicrobial defense. We investigated the effects of
Atsuko Kamo et al.
Journal of dermatological science, 62(2), 91-97 (2011-04-05)
UV-based therapy has anti-pruritic effects in inflammatory skin diseases, such as atopic dermatitis and psoriasis. These anti-pruritic effects may be partly due to inhibition of intraepidermal nerve growth, but they have not been fully characterized. This study was performed to
Prakash Adhikari et al.
International journal of pediatric otorhinolaryngology, 75(4), 500-503 (2011-02-05)
This study was carried out with the objective of comparing clinical efficacy of 10% ichthammol glycerine (IG) pack with steroid-antibiotic pack for relieving pain in cases of acute otitis externa. A prospective quasi-randomized clinical trial was completely carried out in
Christine Bangert et al.
Dermatology (Basel, Switzerland), 222(1), 36-48 (2010-12-15)
Topical pimecrolimus may maintain remissions of atopic dermatitis (AD) by inhibiting subclinical inflammation. To evaluate clinical and cytological effects of pimecrolimus in topical corticosteroid-treated and resolved AD lesions. Patients (n=67) with resolved AD lesions were randomized to 3-week double-blind treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service