Skip to Content
Merck
All Photos(1)

Key Documents

PHR1068

Supelco

Ethylenediaminetetraacetic acid disodium salt dihydrate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Edetate Disodium, Edetate disodium dihydrate, Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Beilstein:
3900609
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. D2900000
traceable to USP 1233009

API family

edta

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

248 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.
Ethylenediaminetetraacetic acid disodium salt dehydrate may be used as a reference standard in validating the potential antioxidant activity and the total phenolic content of samples of seaweed extract via metal chelation technique using ferrous ion-chelating ability assay.
Ethylenediaminetetraacetic acid disodium salt dihydrate may be used as a pharmaceutical reference standard for the determination of ethylenediaminetetraacetic acid (EDTA) in pharmaceutical formulations using ion chromatography with suppressed conductimetric detection.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA5634 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total phenolic compounds, radical scavenging and metal chelation of extracts from Icelandic seaweeds
Wang T, et al.
Food Chemistry, 116(1), 240-248 (2009)
EDTA determination in pharmaceutical formulations and canned foods based on ion chromatography with suppressed conductimetric detection
Krokidis AA, et al.
Analytica Chimica Acta, 535(1-2), 57-63 (2005)
G Lima-Oliveira et al.
British journal of biomedical science, 70(1), 6-9 (2013-04-27)
This study assesses the use of different dry K2 (dipotassium) EDTA vacuum tubes and whether or not they might represent a bias in haematological testing. Blood was collected in three dipotassium EDTA vacuum tubes from different manufacturers: Venosafe, Vacuette and
Gervasio A Lamas et al.
American heart journal, 168(1), 37-44 (2014-06-24)
Disodium ethylenediaminetetraacetic acid (EDTA) reduced adverse cardiac outcomes in a factorial trial also testing oral vitamins. This report describes the intent-to-treat comparison of the 4 factorial groups overall and in patients with diabetes. This was a double-blind, placebo-controlled, 2 ×
Sabrina Lusvarghi et al.
Nucleic acids research, 41(13), 6637-6649 (2013-05-04)
Interaction between the viral protein Rev and the RNA motifs known as Rev response elements (RREs) is required for transport of unspliced and partially spliced human immunodeficiency virus (HIV)-1 and HIV-2 RNAs from the nucleus to the cytoplasm during the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service