Skip to Content
Merck
All Photos(1)

Key Documents

PHR1040

Supelco

Naproxen

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

NAP, (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
Beilstein:
3591068
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 435
traceable to Ph. Eur. N0250000
traceable to USP 1457301

API family

naproxen

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

152-154 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COc1ccc2cc(ccc2c1)[C@H](C)C(O)=O

InChI

1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m0/s1

InChI key

CMWTZPSULFXXJA-VIFPVBQESA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Naproxen belongs to the class of arylacetic acids and is used as a non-steroidal anti-inflammatory drug (NSAID). It is useful for the management of pain, inflammation and stiffness. It is also used in the treatment of rheumatoid arthritis and other painful musculoskeletal disorders.[1][2]

Application

Naproxen may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by high performance liquid chromatography (HPLC) technique.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Non-selective cyclooxygenase (COX-1 and COX-2) inhibitor.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8968 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of naproxen, ketoprofen and phenol red in samples from rat intestinal permeability studies: HPLC method development and validation
Zakeri-Milani P, et al.
Journal of Pharmaceutical and Biomedical Analysis, 39(3-4), 624-630 (2005)
High performance liquid chromatography with photo diode array for separation and analysis of naproxen and esomeprazole in presence of their chiral impurities: enantiomeric purity determination in tablets
Ragab MAA and Eman I
Journal of Chromatography A, 1497, 110-117 (2017)
Jiayang Li et al.
Journal of the American Chemical Society, 135(2), 542-545 (2012-11-10)
As systemically used therapeutics for treating acute or chronic pains or inflammations, nonsteroidal anti-inflammatory drugs (NSAIDs) also associate with the adverse gastrointestinal and renal effects and cardiovascular risks. Thus, it is beneficial to develop topical gels that selectively inhibit cyclooxygenase-2
Michela Campolo et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(11), 4489-4499 (2013-08-01)
Spinal cord trauma causes loss of motor function that is in part due to the ensuing inflammatory response. Hydrogen sulfide (H2S) is a potent, endogenous anti-inflammatory and neuroprotective substance that has been explored for use in the design of novel
Jenny-Maria Brozinski et al.
Environmental science & technology, 47(1), 342-348 (2012-11-29)
Pharmaceutical residues are ubiquitous in rivers, lakes, and at coastal waters affected by discharges from municipal wastewater treatment plants. In this study, the presence of 17 different pharmaceuticals and six different phase I metabolites was determined in the bile of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service