Skip to Content
Merck
All Photos(4)

Key Documents

N1636

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide hydrate

purified by column chromatography, ≥99%

Synonym(s):

β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2 · xH2O
CAS Number:
Molecular Weight:
663.43 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
39200202
PubChem Substance ID:
NACRES:
NA.55

Quality Level

Assay

≥99%

form

crystalline powder

color

white

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2.O[C@@H]3[C@@H](COP(O)(OP(OC[C@@H](O4)[C@@H](O)[C@@H](O)[C@H]4[N+]5=CC=CC(C(N)=O)=C5)([O-])=O)=O)OC[C@@H]3O

InChI

1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)/t10-,11-,13-,14-,15-,16-,20-,21-/m1/s1

InChI key

BAWFJGJZGIEFAR-NNYOXOHSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Nicotinamide adenine dinucleotide (NAD) is an electron carrier and a cofactor. It is ubiquitously found in the living system. NAD+ contains an adenylic acid and a nicotinamide-5′-ribonucleotide group linked together by a pyrophosphate moiety. In NAD+ complexes, the enzyme-cofactor interactions are highly conserved.

Application

β-Nicotinamide adenine dinucleotide hydrate has been used:
  • to measure adenosine triphosphate (ATP) synthesis rates (kinetics measurements)
  • in the reaction buffer for pyruvate kinase (PK) activity assay
  • as a component of master mix for a high throughput screening

Biochem/physiol Actions

β-Nicotinamide adenine dinucleotide (NAD) cycles between the oxidized (NAD+) and reduced (NADH) forms to maintain a redox balance necessary for continued cell growth. It significantly participates in enzyme-catalyzed oxido-reduction processes and many genetic processes. It is also involved in microbial catabolism.
Electron acceptor

Packaging

Packaged by solid weight.

Other Notes

Crystalline NAD contains less than 0.1% (w/w) residual organic solvent, is less hygroscopic and electrostatic, and is more stable at ambient temperatures than amorphous NAD.
This is the common form of NAD.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Techniques to monitor glycolysis
Test, 542(15), 91-114 (2014)
Identification and characterization of novel sirtuin inhibitor scaffolds
Sanders BD, et al.
Bioorganic & Medicinal Chemistry, 17(19), 7031-7041 (2009)
Isolation of Tomato Fruit Chromoplasts and Determination of ATP Levels
Hernandez GF, et al.
Bio-protocol, 4(15), e1192-e1192 (2014)
A Mechanism of Adsorption of beta-Nicotinamide Adenine Dinucleotide on Graphene Sheets: Experiment and Theory
Pumera M, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 15(41), 10851-10856 (2009)
Experimental and theoretical electron density studies in large molecules: NAD+, beta-nicotinamide adenine dinucleotide
Guillot B, et al.
The Journal of Physical Chemistry B, 107(34), 9109-9121 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service