Skip to Content
Merck
All Photos(2)

Key Documents

A7159

Sigma-Aldrich

Bovine Serum Albumin solution

25% in Tyrode's buffer, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27
Pricing and availability is not currently available.

biological source

bovine

Quality Level

sterility

aseptically filled

Assay

23.0-27.0% protein basis (biuret)

form

solution

purified by

heat shock fractionation

does not contain

preservative
stabilizer

origin

USA origin

concentration

23.0-27.0% protein (biuret)
25% in Tyrode's buffer

technique(s)

enzyme immunoassay: suitable

UniProt accession no.

foreign activity

BT virus, none detected
VSV virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin is a globular protein. Bovine serum albumin (BSA) is composed of a single polypeptide chain and cross-linked cysteine disulfide bridges. BSA is heart-shaped and is made up of three homologous domains I-III and each domain has two subdomains A and B.

Biochem/physiol Actions

Albumin acts as a major factor in determining a cow′s health, meat, and milk quality. Serum albumin constitutes about 52-62% of the total water-soluble fraction proteins in the blood plasma. Albumin is a multifunctional protein that plays a role in physiological and pharmacological aspects. It is involved in the transport of fatty acids, bile pigments, cholesterol, drugs, and metals. This protein regulates osmotic pressure and distributes fluids between various compartments.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claus G Madsen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 92, 1-7 (2015-02-11)
This study describes how protein release from polymer matrices correlate with simple measurements on the intrinsic viscosity of the polymer solutions used for casting the matrices and calculations of the solubility parameters of polymers and solvents used. Matrices of poly(dl-lactide-co-glycolide)
Nikolay N Brandt et al.
Journal of biomedical optics, 20(5), 051015-051015 (2014-12-06)
The analysis of the structure-function relationship is extremely important in the study of proteins. The importance of function-related motions of large parts or subglobules of protein molecules stimulates the spectroscopic study in the low-frequency (terahertz) domain. However, only tentative assignments
Christopher L Nobles et al.
Journal of microbiological methods, 118, 7-17 (2015-08-09)
Bacterial pathogens acquire host iron to power cellular processes and replication. Heme, an iron-containing cofactor bound to hemoglobin, is scavenged by bacterial proteins to attain iron. Methods to measure intracellular heme are laborious, involve complex chemistry, or require radioactivity. Such
Gayathri Kanika et al.
Journal of biochemical and molecular toxicology, 29(8), 349-359 (2015-03-17)
Several reports indicated that histone deacetylases (HDACs) play a crucial role in inflammation and fibrogenesis. Sodium butyrate (SB) is a short-chain fatty acid having HDAC inhibition potential. The present study aimed to evaluate the protective effect of SB against L-arginine
Nancy Hachicho et al.
PloS one, 10(8), e0134755-e0134755 (2015-08-21)
Morphology and physiology of fish embryos undergo dramatic changes during their development until the onset of feeding, supplied only by endogenous yolk reserves. For obtaining an insight how these restructuring processes are reflected by body mass related parameters, dry weights

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service