Skip to Content
Merck
All Photos(1)

Key Documents

A8412

Sigma-Aldrich

Bovine Serum Albumin solution

7.5% in DPBS, sterile-filtered, BioXtra, suitable for cell culture

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

biological source

bovine

description

low endotoxin

sterility

sterile-filtered

product line

BioXtra

Assay

7.0-8.0% protein basis (UV)

form

liquid

mol wt

66.4 kDa

purified by

cold ethanol and heatshock fractionation

origin

USA origin

concentration

7.5% in DPBS

technique(s)

cell culture | mammalian: suitable

impurities

≤13.0 EU/mL Endotoxin level

pH

6.5-7.5

foreign activity

Mycoplasma test (Negative)
cell growth >50%

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

BSA is a single polypeptide chain consisting of about 583 amino acid residues and no carbohydrates. At pH 5-7 it contains 17 intrachain disulfide bridges and 1 sulfhydryl group.

Application

Qualified for use in cell culture applications.

Biochem/physiol Actions

Bovine serum albumin is broadly used as an additive to cell culture media, especially serum-free media (SFM). It provides a range of benefits including protection from oxidative damage and stabilization of other media components such as fatty acids and pyridoxal.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda J Shapiro et al.
Veterinary microbiology, 177(1-2), 154-161 (2015-03-18)
The seroprevalence of Coxiella burnetii (C. burnetii) in cats in eastern Australia is unknown, and the risk of transmission from cats to humans is undetermined. This study aimed to determine the exposure of cats to C. burnetii in four distinct
Pinar Sahin et al.
Fertility and sterility, 102(5), 1482-1490 (2014-09-23)
To evaluate the expression of mammalian target of rapamycin (mTOR) pathway molecules in mouse spermatogenesis and as well as its role during proliferation and meiotic initiation of spermatogenic cells. Experimental animal study. University. C57Balb-C adult male mice. Expressions of mTOR
Eric M Kernfeld et al.
Immunity, 48(6), 1258-1270 (2018-06-10)
Thymus development is critical to the adaptive immune system, yet a comprehensive transcriptional framework capturing thymus organogenesis at single-cell resolution is still needed. We applied single-cell RNA sequencing (RNA-seq) to capture 8 days of thymus development, perturbations of T cell receptor rearrangement
Xiyuan Yao et al.
Scientific reports, 10(1), 15201-15201 (2020-09-18)
Cytotoxic drugs that are mechanistically distinct from current chemotherapies are attractive components of personalized combination regimens for combatting aggressive forms of cancer. To gain insight into the cellular mechanism of a potent platinum-acridine anticancer agent (compound 1), a correlation analysis
Katherine Shortt et al.
Scientific reports, 9(1), 1396-1396 (2019-02-06)
Acetaminophen (APAP) is a commonly used analgesic responsible for more than half of acute liver failure cases. Identification of previously unknown genetic risk factors would provide mechanistic insights and novel therapeutic targets for APAP-induced liver injury. This study used a

Articles

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service