Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Activated Charcoal Carbon Adsorbent
Synonym(s):
Charcoal activated, Charcoal activated Norit
®
Empirical Formula (Hill Notation):
C
CAS Number:
7440-44-0
Molecular Weight:
12.01
EC Number:
231-153-3
Products
Product Category
resins (3)
Brand
Supelco (2)
Sustainability
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Separation Technique
Diameter (mm)
Quality Segment
Application
Particle Size (µm)
Special Grade
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
31616
Activated Charcoal Carbon Adsorbent,
puriss. p.a., powder
Expand
View Pricing
29238
Activated Charcoal Carbon Adsorbent,
Norit RBAA-3, 6-7 mesh, rod
Expand
View Pricing
80485
Activated Charcoal Carbon Adsorbent,
for the determination of AOX, 100-285 mesh
Expand
View Pricing