Skip to Content
Merck
All Photos(2)

Key Documents

29238

Supelco

Activated Charcoal Carbon Adsorbent

greener alternative

Norit RBAA-3, 6-7 mesh, rod

Synonym(s):

Charcoal activated Norit®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Beilstein:
4360473
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.54

Product Name

Activated Charcoal Norit®, Norit® RBAA-3, rod

form

rod

Quality Level

manufacturer/tradename

Norit® RBAA-3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

LPLC: suitable

diam.

~3 mm

loss

17-26% loss on drying

matrix

Carbon

matrix active group

carbon

capacity

≥2.1 meq/g neutralization (acid)

greener alternative category

separation technique

reversed phase

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated Charcoal Norit® (Norit RBAA-3, rod) can be used as an adsorbent in the removal of SO2 and NO2 contaminants.

Legal Information

Norit is a registered trademark of Norit N.V.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative studies of low concentration SO2 and NO2 sorption by activated carbon supported [C2mim][Ac] and KOH sorbents
Severa G, et al.
Journal of Environmental Chemical Engineering, 6(1), 718-727 (2018)
Adsorption of 2, 4, 6-trichlorophenol (TCP) onto activated carbon.
Krishnaiah D, et al.
Journal of King Saud University - Science, 25(3), 251-255 (2013)
Harry Marsh
Activated Carbon, Harry Marsh null
Diazepam stability in wastewater and removal by advanced membranes technology, activated carbon and micelle-clay complex
Bufo SA, et al.
Desalination and Water Treatment (2015)
Lithium recovery from aqueous solution by sorption/desorption
Lemaire J, et al.
Hydrometallurgy, 143, 1-11 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service