Skip to Content
Merck
All Photos(1)

Key Documents

05105

Supelco

Activated charcoal

Activated charcoal

powder

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor pressure

<0.1 mmHg ( 20 °C)

Quality Level

description

iodine-adsorption (0.05 mol I2/I) > 70 mL/g
methylene blue-adsorption > 12 mL/0.1 g 0.15% sol.

form

powder

autoignition temp.

842 °F

resistivity

1375 μΩ-cm, 20°C (graphite)

technique(s)

photometry: suitable

ign. residue

≤2%

mp

3550 °C (lit.)

anion traces

chloride (Cl-): ≤5000 mg/kg
sulfate (SO42-): ≤5000 mg/kg

cation traces

Ca: ≤1000 mg/kg
Cd: ≤10 mg/kg
Co: ≤10 mg/kg
Cr: ≤10 mg/kg
Cu: ≤10 mg/kg
Fe: ≤300 mg/kg
K: ≤200 mg/kg
Mg: ≤200 mg/kg
Mn: ≤50 mg/kg
Na: ≤500 mg/kg
Ni: ≤10 mg/kg
Pb: ≤10 mg/kg
Zn: ≤10 mg/kg

application(s)

food and beverages
general analytical

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Activated charcoal can be used for the following:
  • Fiber coating to compare solid-phase microextraction efficiency of the multiwalled carbon nanotubes in water and milk samples before GC-ECD analysis.[1]
  • It may be used to study its effect on tissue cultures of Daucus carota, Happlopapus gracilis and Allum cepa var. proliferum, to check the adsorption of metabolites inhibiting morphognesis.[2]
  • It may also be used in the plant tissue culture to increase the adsorption of auxin and cytokinin from the culture media.[3]
  • It may also be used as the primary gastrointestinal decontamination procedure after acute drug overdose.[4]
  • It may also be used for the adsorption of dyes namely malachite green, bromophenol blue, alizarine red-S, methylene blue, eriochrome black-T, phenol red, methyl blue, and methyl violet from aqueous media.[5]
  • Oral activated charcoal may be also used for adsorption of theophylline, hence can be effectively used in patients with theophylline poisoning.[6]
Activated charcoal or activated carbon is an adsorbent used to remove broad spectrum of pollutants in air, water and soil, and is non-toxic in nature.[7][8] It is odourless and tasteless and can be prepared from any carbonaceous material using gas or chemical activation methods. Gas activation is preferred when the raw material has less than 25% moisture content.[8]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of activated charcoal on tissue cultures: adsorption of metabolites inhibiting morphogenesis
Physiologia Plantarum, 43 (2), 104-106 (1978)
Some effects of activated charcoal as an additive to plant tissue culture media
Weatherhead, M. A., J. Burdon, and G. G. Henshaw.
Journal of Plant Physiology, 89.2 , 141-147 (1978)
Activated charcoal; preparation, characterization and applications: a review article.
Mohammad-Khah, A., and R. Ansari
International Journal of ChemTech Research, 2009, 2745-2788 null
Activated Charcoal: Antidote, Remedy and Health Aid
David O.Cooney
Science, 102-102 (2016)
Muhammad J Iqbal et al.
Journal of hazardous materials, 139(1), 57-66 (2006-07-20)
Adsorption of industrially important dyes namely bromophenol blue, alizarine red-S, methyl blue, methylene blue, eriochrome black-T, malachite green, phenol red and methyl violet from aqueous media on activated charcoal has been investigated. The effect of shaking time, pH and temperature

Questions

1–5 of 5 Questions  
  1. Bonjour, Je prends contact avec vous afin de savoir si ce produit (05105-250G) pourrait être utilisé dans la fabrication de "Sylgard Dishes" (fond noir) pour la dissection de précision et le marquage de tissu ? Je vous remercie

    1 answer
    1. This product is not tested for any specific application, but is expected to be suitable for the preparation of "Sylgard Dishes". Please see the links below to additional components that may be useful in the preparation of these dishes:
      Sylgard 184
      https://www.sigmaaldrich.com/product/aldrich/761036
      Glass Petri dishes
      https://www.sigmaaldrich.com/search/glass-petri-dish?facet=facet_product_category%3Adishes&focus=products&page=1&perpage=30&sort=relevance&term=glass%20petri%20dish&type=product
      Plastic Beakers
      https://www.sigmaaldrich.com/search/plastic-beaker?focus=products&page=1&perpage=30&sort=relevance&term=plastic%20beaker&type=product

      Helpful?

  2. I would like to know what is the electrical conductivity of activated charcoal with 100 mesh particle size. Thank you.

    1 answer
    1. The electrical conductivity of this product was not determined. Please see the link below to review a publication that may be helpful.
      https://link.springer.com/article/10.1007/s13369-014-1516-6

      Helpful?

  3. I would like to know the mesh of this activated carbon charcoal. Thanks a lot

    1 answer
    1. The particle size is not tested internally (and therefore, guaranteed), but the product's supplier sieves the product and claims that 35% or less (by mass) falls through a 40 micron sieve, meaning that at least 65% of the material is smaller than 40 micron. An average particle size is not tested or reported.

      Helpful?

  4. What is the surface area of this activated carbon?

    1 answer
    1. The total surface area for this product is 1050-1150 m²/g.

      Helpful?

  5. I would like to consult about activated charcoal. I want to buy activated carbon powder with a purity greater than 99% and a particle size below 400 mesh (38 microns). If your company offers a suitable product, please contact me.

    1 answer
    1. Unfortunately, there is no activated charcoal that is tested for purity greater than 99% offered. However, there are several products with a particle size smaller than 400 mesh (38 microns). Please see this link for all possible products: https://www.sigmaaldrich.com/search/7440-44-0?focus=products&page=1&perpage=30&sort=relevance&term=7440-44-0&type=cas_number

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service