Skip to Content
Merck
All Photos(1)

Key Documents

68279

Supelco

Sulfuric acid concentrate

0.1 M H2SO4 in water (0.2N), eluent concentrate for IC

Synonym(s):

Sulfuric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NB.21

Quality Level

form

liquid

concentration

0.1 M H2SO4 in water (0.2N)

technique(s)

ion chromatography: suitable

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfuric acid (hydrogen sulfate, sulphine acid,battery acid) is a clear, colorless, oily liquid and is very corrosive in nature towards both metal and tissues. It is soluble in water.
This eluent concentrate for ion chromatography is determined by potentiometric titration. Content and expiry date can be found on the certificate.

Application

Metrohm IC application note AN-S-344:Anions in "Electronic grade" nitric acid on a high-capacity column.

Metrohm IC application note AN-S-341:4-Hydroxybutyratein addition to standard anions and organic acids.

Metrohm IC application note AN-S-336:Anion traces on the Metrosep A Supp 16 - 250/2.0 after Inline Preconcentration and Matrix Elimination

Biochem/physiol Actions

It finds application in the manufacture of explosives, fertilizers, glue and other acids. It is also used in the pickling of metal, purification of petroleum, and in lead-acid batteries. It is widely used as dehydrating agent, catalyst, and as an active reactant in chemical industry. It is also suitable to control pH, ranging from saline solutions to strong fuming acids.
Sulfuric acid is prepared by dry distillation of minerals like copper sulfate pentahydrate (blue vitriol), and iron sulfate heptahydrate (green vitriol). On application of heat these compounds decompose to form iron and copper oxides thereby eliminating water and sulphur trioxide, which then combine to give, dilute solution of sulphuric acid.

Linkage

Visit the IC Portal to learn more

Preparation Note

Prepared with sulfuric acid and high purity water (18.2 MΩ, 0.2 μm filtered)

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Navin G. Ashar, Kiran R. Golwalkar
A Practical Guide to the Manufacture of Sulfuric Acid, Oleums, and Sulfonating Agents (2013)
I A Panagiotopoulos et al.
Bioresource technology, 130, 570-577 (2013-01-22)
A two-stage pretreatment approach, employing steam followed by organosolv treatment, was assessed for its ability to fractionate and recover most of the hemicellulose, lignin and cellulose components of poplar wood chips. A mild steaming stage was initially used to maximise
Francesco Riccobono et al.
Science (New York, N.Y.), 344(6185), 717-721 (2014-05-17)
Atmospheric new-particle formation affects climate and is one of the least understood atmospheric aerosol processes. The complexity and variability of the atmosphere has hindered elucidation of the fundamental mechanism of new-particle formation from gaseous precursors. We show, in experiments performed
João Almeida et al.
Nature, 502(7471), 359-363 (2013-10-08)
Nucleation of aerosol particles from trace atmospheric vapours is thought to provide up to half of global cloud condensation nuclei. Aerosols can cause a net cooling of climate by scattering sunlight and by leading to smaller but more numerous cloud
Ban Xiong Tan et al.
Oncotarget, 5(4), 933-943 (2014-03-25)
Although p53 is found mutated in almost 50% of all cancers, p53 mutations in leukaemia are relatively rare. Acute myeloid leukaemia (AML) cells employ other strategies to inactivate their wild type p53 (WTp53), like the overexpression of the p53 negative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service