Skip to Content
Merck
All Photos(2)

Key Documents

1.09072

Supelco

Sulfuric acid solution

c(H₂SO₄) = 0.5 mol/l (1 N), reag. Ph. Eur., reag. USP, ready-to-use volumetric solution for titration, Titripur®

Synonym(s):

Sulfuric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

Product Name

Sulfuric acid, c(H2SO4) = 0.5 mol/l (1 N), Titripur®, reag. Ph. Eur., reag. USP

Agency

reag. Ph. Eur.
reag. USP

Quality Level

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Acid-base reactions

packaging

Titripac® of

concentration

0.5 M

technique(s)

titration: suitable

pH

0.6 (20 °C in H2O)

density

1.03 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis, characterization, and cytotoxicity studies of nanocellulose extracted from okra (Abelmoschus Esculentus) fiber.: This study uses sulfuric acid for the synthesis and characterization of nanocellulose from okra fiber, evaluating its potential in biomedical applications (Hossen et al., 2024).

  • Synthesis and characterization of mesoporous V-Mo-MCM-41 nanocatalysts: Enhancing efficiency in oxalic acid synthesis.: Explores the use of sulfuric acid in synthesizing V-Mo-MCM-41 nanocatalysts, improving their efficiency in oxalic acid synthesis (Suliman Al Ebraheem et al., 2024).

  • Studies on (polytrimethylene terephthalate)/graphene oxide/f-MWCNT hybrid nanocomposites.: Investigates the preparation of hybrid nanocomposites using sulfuric acid, highlighting their enhanced mechanical and thermal properties (Rath et al., 2024).

  • Probing the dynamics and bottleneck of the key atmospheric SO(2) oxidation reaction by the hydroxyl radical.: Examines the role of sulfuric acid in the atmospheric oxidation of SO₂, providing insights into air quality and climate change (Yuan et al., 2024).

  • Preparation of Bio-Based Polyurethane Coating from Citrullus colocynthis Seed Oil: Characterization and Corrosion Performance.: This research uses sulfuric acid in the synthesis of bio-based polyurethane coatings, assessing their corrosion resistance and environmental benefits (Alshabebi et al., 2024).

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Titripac® packaging - all SKU-pack size numbers ending "4000, 4003, 9010, 9013" - more information on www.sigmaaldrich.com/Titripac

Linkage

Replaces: SX1243K-6

Analysis Note

Form liquid
Amount-of-substance concentration 0.4975 - 0.5025 mol/L
Measurement uncertainty ± 0.0015 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 723 Tris(hydroxymethyl)aminomethane) by means of volumetric standard Tris(hydroxymethyl)aminomethane (article number 1.02408), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Chlorine dioxide acc. to DIN EN ISO 7393

Photometric determination of phosphate ions in manure samples using phosphorus molybdenum blue and Spectroquant® test kits.

Photometric determination of phosphate ions in fertilizer samples using phosphorus molybdenum blue and Spectroquant® test kits.

Photometric determination of phosphate ions in fertilizer samples using phosphorus molybdenum blue and Spectroquant® test kits.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service