Skip to Content
Merck
All Photos(1)

Key Documents

02321

Supelco

2,6-Pyridinedicarboxylic acid

suitable for ion chromatography, ≥99.5% (T)

Synonym(s):

DPA, DPAc, Dipicolinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO4
CAS Number:
Molecular Weight:
167.12
Beilstein:
131629
EC Number:
MDL number:
UNSPSC Code:
41116105
eCl@ss:
39151816
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.5% (T)

technique(s)

ion chromatography: suitable

mp

248-250 °C (dec.) (lit.)

solubility

H2O: 1%, clear

anion traces

phosphate (PO43-): ≤5 mg/kg

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
K: ≤100 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

OC(=O)c1cccc(n1)C(O)=O

InChI

1S/C7H5NO4/c9-6(10)4-2-1-3-5(8-4)7(11)12/h1-3H,(H,9,10)(H,11,12)

InChI key

WJJMNDUMQPNECX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Metrohm IC application note AN-C-163: Cations inbrine with minimal dilution and sub-μL injection.

Metrohm IC application note AN-C-172: Cations insnow from an open field.

Metrohm IC application note AN-C-143: Ammonia inaddition to standard cations in maritime pore water.

Linkage

Visit the IC Portal to learn more

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

370.4 °F - closed cup

Flash Point(C)

188 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Hsiang Lee et al.
Pathogens (Basel, Switzerland), 9(12) (2020-12-04)
Hypermucoviscosity phenotypic Klebsiella pneumoniae (HV-Kp) serotype K1 is the predominant pathogen of a pyogenic liver abscess, an emerging infectious disease that often complicates septic metastatic syndrome in diabetic patients with poor sugar control. HV-Kpisolates were more resistant to neutrophil phagocytosis
Lingbo Kong et al.
Journal of biomedical optics, 19(1), 011003-011003 (2013-07-12)
The dynamics of bacterial spore germination were successfully observed using a fast Raman imaging system, in combination with real-time phase contrast microscopy. By using a multifocus scan scheme, the spontaneous Raman-scattering imaging acquisition speed was increased to ~30 s per
P R Sajanlal et al.
Nanoscale, 4(11), 3427-3437 (2012-04-27)
We present the synthesis of highly anisotropic nickel nanowires (NWs) and large area, free-standing carpets extending over cm(2) area by simple solution phase chemistry. The materials can be post-synthetically manipulated to produce hybrid tubes, wires, and carpets by galvanic exchange
Hui Xu et al.
Chemical communications (Cambridge, England), 48(59), 7377-7379 (2012-05-26)
A nanoscale MOF material NMOF 1 with controllable morphologies is realized whose morphology control has been simulated based on the BFDH method. The targeted NMOF 1 exhibits highly sensitive, selective and instant "turn-on" sensing of bacterial endospores.
Bente Aa Lomstein et al.
Nature, 484(7392), 101-104 (2012-03-20)
Two decades of scientific ocean drilling have demonstrated widespread microbial life in deep sub-seafloor sediment, and surprisingly high microbial-cell numbers. Despite the ubiquity of life in the deep biosphere, the large community sizes and the low energy fluxes in this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service