Skip to Content
Merck
All Photos(1)

Key Documents

36486

Supelco

Sodium bicarbonate concentrate

0.1 M NaHCO3 in water, eluent concentrate for IC

Synonym(s):

Sodium bicarbonate solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NB.21

Quality Level

form

liquid

concentration

0.1 M NaHCO3 in water

technique(s)

ion chromatography: suitable

SMILES string

[Na+].OC([O-])=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

UIIMBOGNXHQVGW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

The “bicarbonate concentrate” is basically 0.1-molar solution of sodium bicarbonate (the saturation temperature of the 0.1-molar solution is 15°C). It is highly advantageous as it is temperature dependent and its subsequent dilution with water can be appropriately controlled.
This eluent concentrate for ion chromatography is determined by potentiometric titration. Content and expiry date can be found on the certificate.

Application

Metrohm IC application note AN-S-344:Anions in "Electronic grade" nitric acid on a high-capacity column.

Metrohm IC application note AN-S-341:4-Hydroxybutyratein addition to standard anions and organic acids.

Metrohm IC application note AN-S-336:Anion traces on the Metrosep A Supp 16 - 250/2.0 after Inline Preconcentration and Matrix Elimination

Linkage

Visit the IC Portal to learn more

Preparation Note

Prepared with sodium bicarbonate and high purity water (18.2 MΩ, 0.2 μm filtered)

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cardiovascular Disorders in Hemodialysis.
Ronco C
Science, 20-22 (2005)
Nimrit Goraya et al.
Current opinion in nephrology and hypertension, 22(2), 193-197 (2013-02-06)
Most patients with chronic kidney disease (CKD) have progressive decline in glomerular filtration rate (GFR), despite current treatment practices. Recent studies support that dietary acid reduction with oral sodium based alkali or base-inducing food types add kidney protection to that
Sumita Agrawal et al.
The Journal of the Association of Physicians of India, 60, 55-56 (2013-02-15)
We report a rare case of a 38-year-old female who presented with sudden onset flaccid quadriplegia and respiratory arrest with no significant past clinical history. She was later found to have hypokalemia due to distal renal tubular acidosis and further
Yepeng Sun et al.
PloS one, 8(1), e53632-e53632 (2013-02-01)
Leymus chinensis (Trin.) Tzvel. is a high saline-alkaline tolerant forage grass genus of the tribe Gramineae family, which also plays an important role in protection of natural environment. To date, little is known about the saline-alkaline tolerance of L. chinensis
James J Barber et al.
Journal of strength and conditioning research, 27(1), 252-258 (2012-12-21)
Creatine and sodium bicarbonate supplementation independently increase exercise performance, but it remains unclear whether combining these 2 supplements is more beneficial on exercise performance. The purpose of this study was to evaluate the impact of combining creatine monohydrate and sodium

Articles

Explore the tests integral to the instrument qualification process for ion chromatography instruments. Learn about the effectiveness of a pre-packaged IQ/OQ/PQ kit, featuring six certified bromide standard solutions for comprehensive IC instrument qualification tests.

Explore the tests integral to the instrument qualification process for ion chromatography instruments. Learn about the effectiveness of a pre-packaged IQ/OQ/PQ kit, featuring six certified bromide standard solutions for comprehensive IC instrument qualification tests.

Explore the tests integral to the instrument qualification process for ion chromatography instruments. Learn about the effectiveness of a pre-packaged IQ/OQ/PQ kit, featuring six certified bromide standard solutions for comprehensive IC instrument qualification tests.

Explore the tests integral to the instrument qualification process for ion chromatography instruments. Learn about the effectiveness of a pre-packaged IQ/OQ/PQ kit, featuring six certified bromide standard solutions for comprehensive IC instrument qualification tests.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service