Skip to Content
Merck
All Photos(4)

Key Documents

56781

Sigma-Aldrich

Iminodiacetic acid

≥98.0% (T), for peptide synthesis

Synonym(s):

IDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2COOH)2
CAS Number:
Molecular Weight:
133.10
Beilstein:
878499
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Product Name

Iminodiacetic acid, purum, ≥98.0% (T)

grade

purum

Quality Level

Assay

≥98.0% (T)

form

solid

mp

243 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)CNCC(O)=O

InChI

1S/C4H7NO4/c6-3(7)1-5-2-4(8)9/h5H,1-2H2,(H,6,7)(H,8,9)

InChI key

NBZBKCUXIYYUSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Iminodiacetic acid (IDA) can be used as a tridentate ligand to chelate metal ions. Some of the common applications of IDA include:
  • Synthesis of N-methyliminodiacetic acid (MIDA), which is an essential precursor for the synthesis of highly versatile MIDA boronates for cross coupling reactions.
  • Synthesis of metal complexes of iminodiacetic acid with a radionuclide, especially 99mTc are as radiotracers for cholescintigraphy scans and for radiopharmaceutical applications.
  • Synthesis of lanthanide hybrid frameworks via self-assembly of lanthanide ions (Ln3+) with iminodiacetic acid under hydrothermal conditions.
  • Preparation of a thermosensitive macroporous protein-imprinted hydrogel for the recognition of protein by metal coordinate interaction.
  • Synthesis of IDA functionalized styrene-divinyl benzene co-polymeric beads for the removal of heavy metal ions like Cd(II), Cr(VI), Ni(II) and Pb(II) from their aqueous solutions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Macroporous thermosensitive imprinted hydrogel for recognition of protein by metal coordinate interaction.
Qin L.
Analytical Chemistry, 81(17), 7206-7216 (2009)
Syntheses, structure, and properties of a series of three-dimensional lanthanide-based hybrid frameworks.
Zhang L.
Inorgorganica Chimica Acta, 400, 67-73 (2013)
General method for synthesis of 2-heterocyclic N-methyliminodiacetic acid boronates.
Dick GR.
Organic Letters, 12(10), 2314-2317 (2010)
Der chelateffekt.
Schwarzenbach VG.
Helvetica Chimica Acta, 35(7), 2344-2359 (1952)
Iminodiacetic acid functionalized cation exchange resin for adsorptive removal of Cr (VI), Cd (II), Ni (II) and Pb (II) from their aqueous solutions.
Misra RK.
Journal of Hazardous Materials, 185(2-3), 1508-1512 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service