Skip to Content
Merck
All Photos(3)

Key Documents

11009

Sigma-Aldrich

L-Arginine

BioUltra, ≥99.5% (NT)

Synonym(s):

(S)-2-Amino-5-guanidinopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
174.20
Beilstein:
1725413
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product line

BioUltra

Quality Level

Assay

≥99.5% (NT)

form

powder or crystals

optical activity

[α]20/D +27.0±0.5°, c = 5% in 5 M HCl

impurities

insoluble matter, passes filter test
≤0.3% foreign amino acids

ign. residue

≤0.05% (as SO4)

loss

≤0.1% loss on drying, 20 °C (HV)

color

white

pH

10.5-12.0 (25 °C, 0.5 M in H2O)

mp

222 °C (dec.) (lit.)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.2
λ: 280 nm Amax: ≤0.1

application(s)

peptide synthesis

SMILES string

N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1

InChI key

ODKSFYDXXFIFQN-BYPYZUCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application


  • L-Arginine-Dependent Nitric Oxide Production in the Blood of Patients with Type 2 Diabetes: A Pilot, Five-Year Prospective Study.: This study examines the role of L-Arginine in nitric oxide production in diabetic patients, proposing its potential benefits for managing vascular complications in diabetes (Stoian et al., 2024).

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Other Notes

Exhibits stabilizing effects on plant protoplasts; pH drift correction in IEF of proteins; Growth requirement of various microorganisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CRC Handbook of Microbiology, 4, 1-1 (1974)
A.W. Galston et al.
Plant Sci. Lett., 11, 69-69 (1978)
H. Delice et al.
Electrophoresis '79, 165-165 (1980)
Krishnan Suresh Kumar et al.
European journal of medicinal chemistry, 45(11), 5474-5479 (2010-08-21)
A new series of 3-(benzylideneamino)-2-phenylquinazoline-4(3H)-ones were prepared through Schiff base formation of 3-amino-2-phenyl quinazoline-4(3)H-one with various substituted carbonyl compounds. Their chemical structures were elucidated by spectral studies. Cytotoxicity and antiviral activity were evaluated against herpes simplex virus-1 (KOS), herpes simplex
Ahmed Moursy et al.
Nucleic acids research, 42(10), 6659-6672 (2014-04-03)
Regulation of SMN2 exon 7 splicing is crucial for the production of active SMN protein and the survival of Spinal Muscular Atrophy (SMA) patients. One of the most efficient activators of exon 7 inclusion is hnRNP G, which is recruited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service