Skip to Content
Merck
All Photos(1)

Key Documents

36544

Supelco

Chloroacetic acid

PESTANAL®, analytical standard

Synonym(s):

Monochloroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COOH
CAS Number:
Molecular Weight:
94.50
Beilstein:
605438
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

3.26 (vs air)

vapor pressure

0.75 mmHg ( 20 °C)

product line

PESTANAL®

Assay

98-100% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

189 °C (lit.)

mp

60-63 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)CCl

InChI

1S/C2H3ClO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

FOCAUTSVDIKZOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroacetic acid belongs to the class of haloacetic acid compounds.[1]
Chloroacetic acid is the degradation product of the herbicide, chloroacetanilide.[2]

Application

Chloroacetic acid may be used as an analytical reference standard for the analysis of chloroacetic acid in:
  • Natural waters by ion chromatography-electrospray ionization-tandem mass spectrometry (IC-ESI-MS/MS).[3]
  • Bottled water, swimming pool water, and both source water and drinking water from a drinking water facility by high performance IC-ESI-MS/MS with multiple reaction monitoring (MRM) detection.[4]
  • Tap water by gas chromatography-mass spectrometry (GC-MS).[5]
  • Drinking water by GC coupled to halogen-specific detector equipped with MS operating under the electron ionization mode.[6]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluating gas chromatography with a halogen-specific detector for the determination of disinfection by-products in drinking water.
Andersson A, et al.
Environmental Science and Pollution Research International, 119(24), 1-10 (2018)
Photodegradation of metolachlor: Isolation, identification, and quantification of monochloroacetic acid
Wilson.IR and Mabury.AS
Journal of Agricultural and Food Chemistry, 48(3), 944-950 (2000)
Determination of bromate and chlorinated haloacetic acids in bottled drinking water with chromatographic methods.
Liu Y and Mou S
Chemosphere, 55(9), 1253-1258 (2004)
Analysis of haloacetic acids, bromate, and dalapon in natural waters by ion chromatography-tandem mass spectrometry.
Wu S, et al.
Journal of Chromatography A, 1487(9), 100-107 (2017)
Rapid simultaneous analysis of 17 haloacetic acids and related halogenated water contaminants by high-performance ion chromatography-tandem mass spectrometry.
Xue R, et al.
Analytical and Bioanalytical Chemistry, 408(24), 6613-6622 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service