Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
174.20
UNSPSC Code:
12352209
NACRES:
NA.26
eCl@ss:
32160406
EC Number:
200-811-1
Beilstein/REAXYS Number:
1725413
MDL number:
biological source
non-animal source
Quality Segment
assay
>98.5%
form
crystalline powder
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, heavy metals, trace metals, residual solvents, tested
mp
222 °C (dec.) (lit.)
solubility
H2O: 100 mg/mL
suitability
suitable for manufacturing use
application(s)
pharmaceutical (small molecule)
SMILES string
N[C@@H](CCCNC(N)=N)C(O)=O
InChI
1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1
InChI key
ODKSFYDXXFIFQN-BYPYZUCNSA-N
Gene Information
human ... NOS1(4842), NOS2(4843)
rat ... Ppm1a(24666)
General description
Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.
M-Clarity Program
Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
M-Clarity Program
Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to the documentation for this product requires a confidentiality disclosure agreement.
Application
L-Arginine is a non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Biochem/physiol Actions
Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.
Legal Information
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under L-Arginine
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.