Skip to Content
Merck
All Photos(1)

Key Documents

46598

Supelco

Oxytetracycline hydrochloride

VETRANAL®, analytical standard

Synonym(s):

5-Hydroxytetracycline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O9 · HCl
CAS Number:
Molecular Weight:
496.89
Beilstein:
3853107
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E703
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

cleaning products
clinical
cosmetics
environmental
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

Mode of action

protein synthesis | interferes

SMILES string

Cl.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O9.ClH/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29;/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31);1H/t12-,13-,14+,17+,21-,22+;/m1./s1

InChI key

UBDNTYUBJLXUNN-IFLJXUKPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Didier Raoult et al.
PLoS pathogens, 4(2), e41-e41 (2008-02-20)
"Nanobacteria" are nanometer-scale spherical and ovoid particles which have spurred one of the biggest controversies in modern microbiology. Their biological nature has been severely challenged by both geologists and microbiologists, with opinions ranging from considering them crystal structures to new
A L Stanton et al.
Journal of dairy science, 95(9), 4950-4960 (2012-08-25)
Bovine respiratory disease complex (BRD) is a common disease in weaned dairy calves that incurs economic and welfare costs. This study was an extension of a randomized clinical trial in which a single injection of tulathromycin (TUL) or oxytetracycline (TET)
Miaomiao Liu et al.
Applied microbiology and biotechnology, 97(19), 8805-8812 (2012-11-29)
To investigate how the microbial community in activated sludge responded to high antibiotic levels, a bench-scale aerobic wastewater treatment system was used to treat oxytetracycline (OTC) mother liquor (OTC-ML). Removal efficiency of chemical oxygen demand decreased from 64.9 to 51.0 %
M Gatti et al.
Theriogenology, 78(8), 1796-1799 (2012-08-29)
Vaginal secretions are an important source of chemical signals, which affect ewes' attractiveness. Moreover, alterations of vaginal flora reduce sexual attractiveness of estrous ewes. As intravaginal sponges containing progestagens (widely used for estrous synchronization) affect vaginal flora, our aims were
Guang-Feng Yang et al.
Bioresource technology, 126, 187-192 (2012-10-18)
A batch test was employed to analyze the joint toxicity of copper (II) and oxytetracycline (OTC), OTC and sulfide, phenol and sulfide (S(2-)), phenol and copper (II), and OTC, copper (II) and substrate on an Anammox mixed culture. The joint

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service