Skip to Content
Merck

46802

Sulfamethazine

VETRANAL®, analytical standard

Synonym(s):

4,6-Dimethylsulfadiazine, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide, Sulfadimethyldiazine, Sulfadimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H14N4O2S
CAS Number:
Molecular Weight:
278.33
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
200-346-4
Beilstein/REAXYS Number:
261304
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

application(s)

clinical testing

format

neat

mode of action

DNA synthesis | interferes, enzyme | inhibits

SMILES string

Cc1cc(C)nc(NS(=O)(=O)c2ccc(N)cc2)n1

InChI

1S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

ASWVTGNCAZCNNR-UHFFFAOYSA-N

General description

Chemical structure: sulfonamide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

An antimicrobial sulfur drug. Induces CYP3A4 expression and acetylated by N-acetyltransferase. Exhibits sex dependent pharmacokinetics, metabolized by the male specific isoform CYP2C11.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Sulfamethazine


Storage Class

11 - Combustible Solids

wgk

WGK 2

ppe

Eyeshields, Gloves, type N95 (US)

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library