Skip to Content
Merck

46161

Demeclocycline hydrochloride hydrate

VETRANAL®, analytical standard

Synonym(s):

Chlorodemethyltetracycline hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H21ClN2O8 · HCl · xH2O
CAS Number:
Molecular Weight:
501.31 (anhydrous basis)
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
200-592-2
Beilstein/REAXYS Number:
5705221
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

O.Cl.CN(C)[C@H]1[C@@H]2C[C@@H]3[C@H](O)c4c(Cl)ccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O

InChI

1S/C21H21ClN2O8.ClH.H2O/c1-24(2)14-7-5-6-10(16(27)12-9(25)4-3-8(22)11(12)15(6)26)18(29)21(7,32)19(30)13(17(14)28)20(23)31;;/h3-4,6-7,14-15,25-26,28-29,32H,5H2,1-2H3,(H2,23,31);1H;1H2/t6-,7-,14-,15-,21-;;/m0../s1

InChI key

LZJOUJFCRCXBBC-XZJYMHMKSA-N

General description

Chemical structure: tetracycline

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Demeclocycline hydrochloride hydrate may be used as an internal standard for the determination of:
  • Oxytetracycline and tetracycline in pig manure samples by ultra-high performance liquid chromatography (UHPLC) coupled to triple quadrupole mass spectrometer equipped with electrospray ionization source (QqQ-ESI-MS/MS) and dynamic multiple reaction monitoring (dMRM) mode of detection.
  • Oxytetracycline hydrochloride in dairy products by UHPLC-QqQ-ESI-MS/MS.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library