Skip to Content
Merck
All Photos(2)

Documents

31626

Supelco

Bacitracin A

VETRANAL®, analytical standard, main stereoisomer ~50 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H103N17O16S
Molecular Weight:
1422.69
EC Number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E700
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

composition

main stereoisomer, ~50%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC[C@H](C)[C@H](N)C1=N[C@@H](CS1)C(=O)N[C@@H](CC(C)C)C(=O)N[C@H](CCC(O)=O)C(=O)NC([C@@H](C)CC)C(=O)N[C@H]2CCCCNC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](CC(O)=O)NC(=O)[C@H](Cc3cnc[nH]3)NC(=O)[C@@H](Cc4ccccc4)NC(=O)C(NC(=O)[C@@H](CCCN)NC2=O)[C@@H](C)CC

Looking for similar products? Visit Product Comparison Guide

General description

Bacitracin is a polypeptide antibiotic produced by strains of Bacillus subtilis and Bacillus licheniformis. Bacitracin A is a commercially produced, major microbiologically active component of bacitracin.

Application

Commission Regulation (EU) No 37/2010 of 22 December 2009 on pharmacologically active substances and their classification regarding maximum residue limits in foodstuffs of animal origin
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M Siegel et al.
Biological mass spectrometry, 23(4), 186-204 (1994-04-01)
The bacitracin antibiotic complex consists principally of bacitracin A, a peptide antibiotic containing seven amino acid residues in a ring and five amino acid residues in a blocked side chain, together with a mixture of minor components presumably related but
L W Tetler et al.
Biological mass spectrometry, 22(12), 712-720 (1993-12-01)
Bacitracin is a mixture of polypeptide antibiotics produced by the action of Bacillus subtilis and Bacillus licheniformis. The mixture has previously been shown to contain at least ten components, of which only the major component, Bacitracin A, and its oxidized
Quantitative high-pressure liquid chromatographic analysis of bacitracin, a polypeptide antibiotic.
Tsuji K, et al.
Journal of Chromatography A, 99(4), 597- 608 (1974)
Mukundan Ragavan et al.
Analytical chemistry, 83(15), 6054-6059 (2011-06-10)
Hyperpolarization of nuclear spins through techniques such as dynamic nuclear polarization (DNP) can greatly increase the signal-to-noise ratio in NMR measurements, thus eliminating the need for signal averaging. This enables the study of many dynamic processes which would otherwise not
Walter Mancino et al.
Microorganisms, 7(12) (2019-12-08)
Specific members of the genus Bifidobacterium are among the first colonizers of the human/animal gut, where they act as important intestinal commensals associated with host health. As part of the gut microbiota, bifidobacteria may be exposed to antibiotics, used in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service