Skip to Content
Merck

32161

Gamithromycin

VETRANAL®, analytical standard

Synonym(s):

(2R,3S,4R,5S,8RR,10R,11R,12S,13S,14R)-13-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10-trihydroxy-3,5,8,10,12,14-hexamethyl-7-propyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylohexopyranosyl]oxy]-1-oxa-7-azacyclopentadecan-15-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C40H76N2O12
CAS Number:
Molecular Weight:
777.04
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

technique(s)

HPLC: suitable, chromatography: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

N1([C@@H](C[C@]([C@@H](C([C@@H]([C@H](C(=O)O[C@@H]([C@]([C@@H]([C@H](C1)C)O)(O)C)CC)C)O[C@@H]3O[C@H]([C@@H]([C@@](C3)(OC)C)O)C)C)O[C@@H]2O[C@@H](C[C@@H]([C@H]2O)N(C)C)C)(O)C)C)CCC

InChI

1S/C40H76N2O12/c1-15-17-42-21-22(3)33(44)40(11,48)29(16-2)52-36(46)26(7)32(53-30-20-39(10,49-14)34(45)27(8)51-30)25(6)35(38(9,47)19-23(42)4)54-37-31(43)28(41(12)13)18-24(5)50-37/h22-35,37,43-45,47-48H,15-21H2,1-14H3/t22-,23+,24+,25?,26+,27-,28-,29+,30-,31+,32-,33+,34-,35+,37-,38+,39+,40+/m0/s1

InChI key

VWAMTBXLZPEDQO-FWEXOAGLSA-N

General description

Gamithromycin is a drug, which is administered to cattle for the treatment of bovine respiratory disease caused by Pasteurella multocida, Histophilus somni and Mannheimia haemolytica. It is also effective in treating certain infections caused by Mycoplasma.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Gamithromycin may be used as a reference standard in the determination of gamithromycin in samples of plasma, lung tissue and pulmonary epithelial lining fluid using liquid chromatography coupled with tandem mass spectrometry (LC-MS-MS).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library