Skip to Content
Merck
All Photos(3)

Key Documents

D199303

Sigma-Aldrich

2,4-Dinitrophenylhydrazine

reagent grade, 97%

Synonym(s):

(2,4-dinitrophenyl)-hydrazine, DNPH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3NHNH2
CAS Number:
Molecular Weight:
198.14
Beilstein:
615586
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
Assay:
97%
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

Assay

97%

contains

33% water (minimum)

mp

197-200 °C (lit.)

functional group

hydrazine
nitro

SMILES string

NNc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H6N4O4/c7-8-5-2-1-4(9(11)12)3-6(5)10(13)14/h1-3,8H,7H2

InChI key

HORQAOAYAYGIBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dinitrophenylhydrazine (DNPH), a hydrazine derivative, is a potential mutagenic agent. Its molecular and crystal structure has been analyzed.[1] The intra- and intermolecular hydrogen bonds in DNPH have been investigated by vibrational spectroscopic analysis.[2] The mechanism of isothermal degradation of DNPH has been studied by differential scanning calorimetry (DSC).[3]

Application

2,4-Dinitrophenylhydrazine has been used as a reagent to detect the presence of aldehydes and ketones in protein carbonyls.[4][5][6] It may be used as a colorimetric reagent in the quantitative estimation of prednisone.[7]

Packaging

500G contains total wet product weight of ~485G. Variance is due to addition of water to meet DOT requirements.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Desen. Expl. 1

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Chem. Soc. Pak., 14, 73-73 (1992)
Clare L Hawkins et al.
Free radical biology & medicine, 46(8), 965-988 (2009-05-15)
Proteins are major targets for oxidative damage due to their abundance and rapid rates of reaction with a wide range of radicals and excited state species, such as singlet oxygen. Exposure of proteins to these oxidants results in loss of
Structure of 2, 4-dinitrophenylhydrazine.
Okabe N, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 49(9), 1678-1680 (1993)
Journal of the American Oil Chemists' Society, 70, 881-881 (1993)
Kinetic analysis of isothermal decomposition of 2, 4-dinitrophenylhydrazine using differential scanning calorimetry.
Musuc AM, et al.
Thermochimica Acta, 448(2), 130-135 (2006)

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Before use, is it necessary to dry off the water content in this reagent? If so, what would be the temperature and how long without triggering degradation of the reagent?

    1 answer
    1. This product is intentionally provided as a moist powder, with at least 33% water by weight. The material may be safely handled in the moistened state, but not as a dry powder. The recommendation would be to NOT dry off the water content.

      Helpful?

  4. We have this product (D199303-500G) on our site and want to dispose of it . Can you please give us instructions on how to dispose the product safely ?

    1 answer
    1. Please refer to section 13 of the Safety Data Sheet for disposal information. Consult with facility or municipal EHS authorities to assure proper guidelines are followed. See the link below to review the SDS:
      https://www.sigmaaldrich.com/sds/aldrich/d199303

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service